Merck
  • Início
  • Resultados da busca
  • A novel splice variant of Gαq-coupled Bombyx CAPA-PVK receptor 1 functions as a specific Gαi/o-linked receptor for CAPA-PK.
A novel splice variant of Gαq-coupled Bombyx CAPA-PVK receptor 1 functions as a specific Gαi/o-linked receptor for CAPA-PK.
Biochimica et biophysica acta. Molecular cell research (2020-04-15)
Zheng Cao, Lili Yan, Zhangfei Shen, Yu Chen, Ying Shi, Xiaobai He, Naiming Zhou
RESUMO
Alternative splicing enables G protein-coupled receptor (GPCR) genes to greatly increase the number of structurally and functionally distinct receptor isoforms. However, the functional role and relevance of the individual GPCR splice variants in regulating physiological processes are still to be assessed. A naturally occurring alternative splice variant of Bombyx CAPA-PVK receptor, BomCAPA-PVK-R1-Δ341, has been shown to act as a dominant-negative protein to regulate cell surface expression and function of the canonical CAPA-PVK receptor. Herein, using functional assays, we identify the splice variant Δ341 as a specific receptor for neuropeptide CAPA-PK, and upon activation, Δ341 signals to ERK1/2 pathway. Further characterization demonstrates that Δ341 couples to Gαi/o, distinct from the Gαq-coupled canonical CAPA-PVK receptor, triggering ERK1/2 phosphorylation through Gβγ-PI3K-PKCζ signaling cascade. Moreover, our ELISA data show that the ligand-dependent internalization of the splice variant Δ341 is significantly impaired due to lack of GRKs-mediated phosphorylation sites. Our findings highlight the potential of this knowledge for molecular, pharmacological and physiological studies on GPCR splice variants in the future.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Poly-L-lysine hydrobromide, mol wt 70,000-150,000, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
U-73122 hydrate, powder
Sigma-Aldrich
Trypsin-EDTA solution, 0.25%, sterile-filtered, BioReagent, suitable for cell culture, 2.5 g porcine trypsin and 0.2 g EDTA • 4Na per liter of Hanks′ Balanced Salt Solution with phenol red
Sigma-Aldrich
Tyrphostin AG 1478, ≥98%
Sigma-Aldrich
Gö 6983, ≥97%
Sigma-Aldrich
Phosphate buffered saline, 10× concentrate, BioPerformance Certified, suitable for cell culture
Sigma-Aldrich
PP2, ≥98% (HPLC)
Sigma-Aldrich
Hanks′ Balanced Salt solution, Modified, with sodium bicarbonate, without phenol red, calcium chloride and magnesium sulfate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
U0126 monoethanolate, ≥98% (HPLC), powder