Sensing with TRP channels.
Nature chemical biology (2006-01-13)
Thomas Voets, Karel Talavera, Grzegorz Owsianik, Bernd Nilius
RESUMO
Drosophila melanogaster flies carrying the trp (transient receptor potential) mutation are rapidly blinded by bright light, because of the absence of a Ca2+-permeable ion channel in their photoreceptors. The identification of the trp gene and the search for homologs in yeast, flies, worms, zebrafish and mammals has led to the discovery of a large superfamily of related cation channels, named TRP channels. Activation of TRP channels is highly sensitive to a variety of chemical and physical stimuli, allowing them to function as dedicated biological sensors that are essential in processes such as vision, taste, tactile sensation and hearing.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Capsaicin, ≥95%, from Capsicum sp.
Sigma-Aldrich
Cinnamaldehyde, natural, ≥95%, FG
Sigma-Aldrich
2-Aminoethyl diphenylborinate, 97%
Sigma-Aldrich
trans-Cinnamaldehyde, 99%
Sigma-Aldrich
Camphor, 96%
Sigma-Aldrich
L-Menthol, natural, ≥99%, FCC, FG
Supelco
(−)-Menthol, analytical standard
Sigma-Aldrich
Capsaicin, natural
Sigma-Aldrich
Allyl isothiocyanate, contains BHT as stabilizer, 95%
Sigma-Aldrich
(1R)-(+)-Camphor, 98%
Supelco
Allyl isothiocyanate, PESTANAL®, analytical standard
Sigma-Aldrich
L-Menthol, ≥99%, FCC, FG
Sigma-Aldrich
(1R,2S,5R)-(−)-Menthol, ReagentPlus®, 99%
Sigma-Aldrich
Allyl isothiocyanate, ≥95%, FCC
Sigma-Aldrich
(±)-Camphor, purum, synthetic, ≥95.0% (GC)
Sigma-Aldrich
(−)-Menthol, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.0-102.0%
Sigma-Aldrich
trans-Cinnamaldehyde, ≥99%
Sigma-Aldrich
D-Camphor, ≥97%, FG
Sigma-Aldrich
(±)-Camphor, meets analytical specification of Ph. Eur., BP, ≥95% (GC), racemic
Sigma-Aldrich
Piperine, ≥95%, FG
Sigma-Aldrich
trans-Cinnamaldehyde, FCC, FG
Sigma-Aldrich
(±)-Camphor, ≥95.5%
(−)-Menthol, primary reference standard