  • Search Results
  • 20(R)-ginsenoside Rh2, not 20(S), is a selective osteoclastgenesis inhibitor without any cytotoxicity.
20(R)-ginsenoside Rh2, not 20(S), is a selective osteoclastgenesis inhibitor without any cytotoxicity.
Bioorganic & medicinal chemistry letters (2009-05-12)
Jie Liu, Jun Shiono, Kuniyoshi Shimizu, Hongshan Yu, Chunzhi Zhang, Fengxie Jin, Ryuichiro Kondo
ABSTRACT
Increased osteoclastic bone resorption plays a central role in the pathogenesis of many bone diseases, and osteoclast inhibitors are the most widely used treatments for these diseases. Ginsenosides, the main component of ginseng, have been known for their medicinal effects such as anti-inflammatory and anti-proliferative activities. In this study, we investigated the inhibitory effects of ginsenosides (ginsenoside 20(R)-Rh2 and ginsenoside 20(S)-Rh2) on osteoclastgenesis using RAW264 cells in vitro. Only ginsenoside 20(R)-Rh2 showed selective osteoclastgenesis inhibitory activity without any cytotoxicity up to 100 microM. These results implied that the stereochemistry of the hydroxyl group at C-20 may play an important role in selective osteoclastgenesis inhibitory activity.
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g