  • Search Results
  • Antidyslipidemic and antioxidative activities of 8-hydroxyquinoline derived novel keto-enamine Schiffs bases.
Antidyslipidemic and antioxidative activities of 8-hydroxyquinoline derived novel keto-enamine Schiffs bases.
European journal of medicinal chemistry (2008-09-30)
Koneni V Sashidhara, Abdhesh Kumar, Gitika Bhatia, M M Khan, A K Khanna, J K Saxena
ABSTRACT
8-Hydroxyquinoline when subjected to Duff reaction resulted in the formation of unexpected 7-methylaminomethylene-8-oxo-7, 8-dihydroquinoline-5-carbaldehyde 2, which existed in the keto-enamine form, in which the aromaticity of the relevant ring was disrupted, which upon subsequent treatment with various primary amines resulted in its nucleophilic substitution of aliphatic methyl amine. These interesting novel derivatives were evaluated in vitro for their antioxidant and in vivo for their antidyslipidemic and post-heparin lipolytic activities. Compound 6 was found to be most active antidyslipidemic and antioxidative agent in this series, respectively, and thus represent a new class of promising lead.
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, ≥98%
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination