Merck
  • Início
  • Resultados da busca
  • A Potential Role for Stress-Induced Microbial Alterations in IgA-Associated Irritable Bowel Syndrome with Diarrhea.
A Potential Role for Stress-Induced Microbial Alterations in IgA-Associated Irritable Bowel Syndrome with Diarrhea.
Cell reports. Medicine (2020-11-17)
Sunaina Rengarajan, Kathryn A Knoop, Arvind Rengarajan, Jiani N Chai, Jose G Grajales-Reyes, Vijay K Samineni, Emilie V Russler-Germain, Prabha Ranganathan, Alessio Fasano, Gregory S Sayuk, Robert W Gereau, Andrew L Kau, Dan Knights, Purna C Kashyap, Matthew A Ciorba, Rodney D Newberry, Chyi-Song Hsieh
RESUMO
Stress is a known trigger for flares of inflammatory bowel disease (IBD) and irritable bowel syndrome (IBS); however, this process is not well understood. Here, we find that restraint stress in mice leads to signs of diarrhea, fecal dysbiosis, and a barrier defect via the opening of goblet-cell associated passages. Notably, stress increases host immunity to gut bacteria as assessed by immunoglobulin A (IgA)-bound gut bacteria. Stress-induced microbial changes are necessary and sufficient to elicit these effects. Moreover, similar to mice, many diarrhea-predominant IBS (IBS-D) patients from two cohorts display increased antibacterial immunity as assessed by IgA-bound fecal bacteria. This antibacterial IgA response in IBS-D correlates with somatic symptom severity and was distinct from healthy controls or IBD patients. These findings suggest that stress may play an important role in patients with IgA-associated IBS-D by disrupting the intestinal microbial community that alters gastrointestinal function and host immunity to commensal bacteria.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Tropicamide, solid
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
Atropine, ≥99% (TLC), powder
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Tyrphostin AG 1478, ≥98%
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by phenol extraction