GuttaFlow® Bioseal Cytotoxicity Assessment: In Vitro Study.
Molecules (Basel, Switzerland) (2020-09-24)
Inês Ferreira, Mafalda Laranjo, Carlos Miguel Marto, João Casalta-Lopes, Beatriz Serambeque, Ana Cristina Gonçalves, Ana Bela Sarmento-Ribeiro, Eunice Carrilho, Maria Filomena Botelho, Anabela Baptista Paula, Manuel Marques Ferreira
RESUMO
The sealers used for root canal treatment should be biocompatible for the peri-radicular tissues, to evaluate the cytotoxic effects of GuttaFlow® bioseal sealer and to compare them with AH26® epoxy resin. Culture media were conditioned with the GuttaFlow® bioseal and AH26® pellets. MDPC-23 odontoblast cell cultures were treated with conditioned medium and serial dilutions. To evaluate the metabolic activity and cellular viability, the MTT and SRB assays were performed. To determine the production of reactive oxygen species, the DHE and DCF-DA probes were used. Cell cycle and cell-death types were assessed by cytometry, and to evaluate the mineralization capacity, the Alizarin Red S coloration was used. Statistical analysis was performed using analysis of variance (ANOVA) when normality was found and Kruskal-Wallis on the opposite case. For the comparison with normality values, the Student t-test was used. Cells exposed to the GuttaFlow® bioseal conditioned medium maintained high metabolic activities, except at higher concentrations. Likewise, viability was maintained, but a significant decrease was observed after exposure to the highest concentration (p < 0.001), associated with cell death by late apoptosis and necrosis. When cell cultures were exposed to AH26®, metabolic activity was highly compromised, resulting in cell death. An imbalance in the production of peroxides and superoxide anion was observed. GuttaFlow® bioseal showed higher biocompatibility than AH26®.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Fetal Bovine Serum, non-USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Antibiotic Antimycotic Solution (100×), Stabilized, with 10,000 units penicillin, 10 mg streptomycin and 25 μg amphotericin B per mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Calcium chloride, anhydrous, powder, ≥97%
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose and L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Alizarin Red S, certified by the Biological Stain Commission
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)