Merck
  • Início
  • Resultados da busca
  • Identifying potential patient-specific predictors for anterior cruciate ligament reconstruction outcome - a diagnostic in vitro tissue remodeling platform.
Identifying potential patient-specific predictors for anterior cruciate ligament reconstruction outcome - a diagnostic in vitro tissue remodeling platform.
Journal of experimental orthopaedics (2020-07-06)
Marc van Vijven, Bart van Groningen, Joyce N Kimenai, Maria C van der Steen, Marina van Doeselaar, Rob P A Janssen, Keita Ito, Jasper Foolen
RESUMO
Upon anterior cruciate ligament (ACL) rupture, reconstruction is often required, with the hamstring tendon autograft as most widely used treatment. Post-operative autograft remodeling enhances graft rupture risk, which occurs in up to 10% of the patient population, increasing up to 30% of patients aged under 20 years. Therefore, this research aimed to identify potential biological predictors for graft rupture, derived from patient-specific tissue remodeling-related cell properties in an in vitro micro-tissue platform. Hamstring tendon-derived cells were obtained from remnant autograft tissue after ACL reconstructions (36 patients, aged 12-55 years), and seeded in collagen I gels on a micro-tissue platform. Micro-tissue compaction over time - induced by altering the boundary constraints - was monitored. Pro-collagen I expression was assessed using ELISA, and protein expression of tenomodulin and α-smooth muscle actin were measured using Western blot. Expression and activity of matrix metalloproteinase 2 were determined using gelatin zymography. Only micro-tissues corresponding to younger patients occasionally released themselves from the constraining posts. Pro-collagen I expression was significantly higher in younger patients. Differences in α-smooth muscle actin and tenomodulin expression between patients were found, but these were age-independent. Active matrix metalloproteinase 2 expression was slightly more abundant in younger patients. The presented micro-tissue platform exposed patient-specific remodeling-related differences between tendon-derived cells, with the micro-tissues that released from constraining posts and pro-collagen I expression best reflecting the clinical age-dependency of graft rupture. These properties can be the starting point in the quest for potential predictors for identifying individual patients at risk for graft rupture.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, Modified, with 36 mg sodium pyruvate, 50 mg streptomycin sulfate, 100 mg kanamycin monosulfate, 1000 mg glucose/L and CaCl2, liquid, 0.1 μm filtered, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid 2-phosphate sesquimagnesium salt hydrate, ≥95%
Sigma-Aldrich
DL-Dithiothreitol, ≥98% (HPLC), ≥99.0% (titration)
Sigma-Aldrich
Fetal Bovine Serum, non-USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Pluronic® F-127, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
RIPA Buffer