• Início
  • Resultados da busca
  • Identification of Specific Glycosyltransferases Involved in Flavonol Glucoside Biosynthesis in Ginseng Using Integrative Metabolite Profiles, DIA Proteomics, and Phylogenetic Analysis.
Identification of Specific Glycosyltransferases Involved in Flavonol Glucoside Biosynthesis in Ginseng Using Integrative Metabolite Profiles, DIA Proteomics, and Phylogenetic Analysis.
Journal of agricultural and food chemistry (2021-01-30)
Qinggang Yin, Xiaoyan Han, Jingwang Chen, Zongxian Han, Liang Shen, Wei Sun, Shilin Chen
RESUMO
Ginseng contains a variety of flavonol glycosides that possess diverse biological activities; however, scant information of flavonoid glycosylation was reported in ginseng. We found that panasenoside and kaempferol 3-O-glucoside were commonly accumulated along with cultivation years in leaves. In order to explore the procedure of flavonol glycosylation in ginseng, 50 UDP-glycosyltransferases (UGTs) were screened out using differentiated data-independent acquisition (DIA) proteomics and phylogenetic analysis. UGT92A10 and UGT94Q4 were found contributing to the formation of kaempferol 3-O-glucoside. UGT73A18, UGT74T4, and UGT75W1 could catalyze galactosylation of kaempferol 3-O-glucoside. Ser278, Trp335, Gln338, and Val339 were found forming hydrogen bonds with UDP-galactose in UGT75W1 by docking. MeJA induced transcripts of UGT73A18 and UGT74T4 by over fourfold, consistent with the decrease of kaempferol 3-O-glucoside, which indicated that these genes may be related to resisting adversity stress in ginseng. These results highlight the significance of integrative metabolite profiles, proteomics, and phylogenetic analysis for exploring flavonol glycosylation in ginseng.
MATERIAIS
Número do produto
Marca
Descrição do produto
Rutin, phyproof® Reference Substance
Sigma-Aldrich
3-Hydroxyflavone, ≥98%