Merck
  • Início
  • Resultados da busca
  • Autocrine secretion of insulin-like growth factor-I mediates growth hormone-stimulated DNA synthesis and proliferation in primary cultures of adult rat hepatocytes.
Autocrine secretion of insulin-like growth factor-I mediates growth hormone-stimulated DNA synthesis and proliferation in primary cultures of adult rat hepatocytes.
European journal of pharmacology (2020-11-28)
Kazuki Kurihara, Hajime Moteki, Mitsutoshi Kimura, Masahiko Ogihara
RESUMO
The intracellular signaling pathway of growth hormone (GH)-stimulated DNA synthesis and proliferation was investigated in primary cultures of adult rat hepatocytes. DNA synthesis and cell proliferation were detected in hepatocyte parenchymal cells grown in serum-free, defined medium containing GH (100 ng/ml). GH-stimulated hepatocyte DNA synthesis and proliferation were almost completely blocked by TG101209 (10-6 M), a selective Janus kinase (JAK)2 inhibitor, U-73122 (10-6 M), a selective phospholipase C (PLC) inhibitor, and a monoclonal antibody to insulin-like growth factor-I (IGF-I) receptor (100 ng/ml) or anti-secretion agents such as somatostatin (10-6 M) and BAPTA/AM (10-7 M). In addition, blocking monoclonal antibodies to IGF-I, but not transforming growth factor-α, completely inhibited GH-induced hepatocyte DNA synthesis and proliferation. IGF-I levels in the culture medium increased rapidly versus baseline levels within 5 min in response to GH (100 ng/ml), and the maximum IGF-I level (100 pg/ml) was reached 20 min after GH stimulation. Autocrine secretion of IGF-I into the culture medium was inhibited by a growth-inhibitory dose of TG101209, U-73122, somatostatin, or BAPTA/AM. These data indicate that the proliferative mechanism of action of GH is mediated mainly through a GH receptor/JAK2/PLC-stimulated increase in the autocrine secretion of IGF-I by primary cultured hepatocytes, followed by stimulation of the 95 kDa IGF-I receptor tyrosine kinase signaling pathway.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
U-73343
Sigma-Aldrich
Tyrphostin AG 1478, ≥98%