  • Search Results
  • Synthesis, biopharmaceutical characterization, antimicrobial and antioxidant activities of 1-(4'-O-β-D-glucopyranosyloxy-2'-hydroxyphenyl)-3-aryl-propane-1,3-diones.
Synthesis, biopharmaceutical characterization, antimicrobial and antioxidant activities of 1-(4'-O-β-D-glucopyranosyloxy-2'-hydroxyphenyl)-3-aryl-propane-1,3-diones.
European journal of medicinal chemistry (2011-02-23)
Javed Sheikh, Ali Parvez, Harjeet Juneja, Vishwas Ingle, Zahid Chohan, Moulay Youssoufi, Taibi Ben Hadda
ABSTRACT
This research communication is toward the investigation of the antibacterial, antifungal and antioxidant activities of the synthesized compounds 1-(4'-O-β-D-glucopyranosyloxy-2'-hydroxyphenyl)-3-aryl-propane-1,3-diones (5a)-(5h). These compounds have been obtained by the interaction of α-acetobromoglucose with 1-(2',4'-dihydroxyphenyl)-3-aryl-propane-1,3-diones (3a)-(3h) under anhydrous condition and at lower temperature. The structures of these newly synthesized O-β-D-glucopyranosides were established on the basis of chemical, elemental, and spectral analyses. Further, the compounds (5b), (5c), (5d) and (5g) showed potent antibacterial and antifungal activity. A good correlation was obtained between the theoretical predictions of bioavailability using Lipinski's rule-of-five and experimental verification.
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)