• Início
  • Resultados da busca
  • Vitamin D Inhibits IL-22 Production Through a Repressive Vitamin D Response Element in the il22 Promoter.
Vitamin D Inhibits IL-22 Production Through a Repressive Vitamin D Response Element in the il22 Promoter.
Frontiers in immunology (2021-08-20)
Daniel V Lopez, Fatima A H Al-Jaberi, Nkerorema D Damas, Brian T Weinert, Urska Pus, Sara Torres-Rusillo, Anders Woetmann, Niels Ødum, Charlotte M Bonefeld, Martin Kongsbak-Wismann, Carsten Geisler
RESUMO
Th22 cells constitute a recently described CD4+ T cell subset defined by its production of interleukin (IL)-22. The action of IL-22 is mainly restricted to epithelial cells. IL-22 enhances keratinocyte proliferation but inhibits their differentiation and maturation. Dysregulated IL-22 production has been associated to some inflammatory skin diseases such as atopic dermatitis and psoriasis. How IL-22 production is regulated in human T cells is not fully known. In the present study, we identified conditions to generate Th22 cells that do not co-produce IL-17 from naïve human CD4+ T cells. We show that in addition to the transcription factors AhR and RORγt, the active form of vitamin D3 (1,25(OH)2D3) regulates IL-22 production in these cells. By studying T cells with a mutated vitamin D receptor (VDR), we demonstrate that the 1,25(OH)2D3-induced inhibition of il22 gene transcription is dependent on the transcriptional activity of the VDR in the T cells. Finally, we identified a vitamin D response element (VDRE) in the il22 promoter and demonstrate that 1,25(OH)2D3-VDR directly inhibits IL-22 production via this repressive VDRE.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
TRI Reagent®, For processing tissues, cells cultured in monolayer or cell pellets
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Ionomycin calcium salt from Streptomyces conglobatus, powder, ≥98% (HPLC)
SAFC
RPMI-1640 Medium, HEPES Modification, With 25 mM HEPES, without L-glutamine., liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Monensin sodium salt, 90-95% (TLC)