Transcriptional IL-15-directed in vivo DC targeting DNA vaccine.
Gene therapy (2009-09-04)
S Tian, Z Liu, C Donahue, H S Noh, L D Falo, Z You
RESUMO
Dendritic cells (DC) engineered in vitro by DNA encoding OVAhsp70 and IL-15 up-regulated their expressions of CD80, CD86, CCR7 and IL-15Ralpha and promoted their productions of IL-6, IL-12 and TNF-alpha. Transcriptional IL-15-directed in vivo DC targeting DNA vaccine encoding OVAhsp70 elicited long-lasting Th1 and CTL responses and anti-B16OVA activity. CD8T cell-mediated primary tumor protection was abrogated by DC or CD4T cell depletion during the induction phase of immune responses. However, CD4T cell depletion during immunization did not impair CD8T cell-dependent long-lasting tumor protection. Furthermore, in vivo DC-derived IL-15 exerted the enhancements of cellular and humoral immune responses and antitumor immunity elicited by OVAhsp70 DNA vaccine. Importantly, the potency of this novel DNA vaccine strategy was proven using a self/tumor Ag (TRP2) in a clinically relevant B16 melanoma model. These findings have implications for developing next generation DNA vaccines against cancers and infectious diseases in both healthy and CD4 deficient individuals.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Antibiotic Antimycotic Solution (100×), Stabilized, with 10,000 units penicillin, 10 mg streptomycin and 25 μg amphotericin B per mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Diphtheria Toxin from Corynebacterium diphtheriae, lyophilized powder, Product is in unnicked form