Merck
Therapeutic effect of magnesium sulphate on carbon monoxide toxicity-mediated brain lipid peroxidation.
European review for medical and pharmacological sciences (2013-04-26)
Y Yavuz, H Mollaoglu, Y Yürümez, K Ucok, L Duran, K Tünay, L Akgün
RESUMO
Carbon monoxide (CO) toxicity primarily results from cellular hypoxia caused by impedance of oxygen delivery. Studies show that CO may cause brain lipid peroxidation and leukocyte-mediated inflammatory changes in the brain. The aim of this study was to investigate whether magnesium sulphate could prevent or diminish brain lipid peroxidation caused by carbon monoxide toxicity in rats. Fourty rats were divided into five groups of 8 rats each. Group l was not received any agent during the experiment. Group 2 was inhaled CO gas followed by intraperitoneally normal saline 30 minutes (min) later. Group 3 was inhaled CO gas followed by 100 mg/kg magnesium sulphate intraperitoneally 30 min later. Group 2 and Group 3 rats was undergone laparotomy and craniotomy while still under anesthesia at 6 hour, and tissue sample was obtained from the cerebrum. Group 4 was inhaled CO gas followed by intraperitoneally normal saline 30 min later. Group 5 was inhaled CO gas followed by 100 mg/kg magnesium sulphate intraperitoneally 30 min later. Group 4 and Group 5 rats was undergone laparotomy and craniotomy while still under anesthesia at 24 hour, and tissue sample was obtained from the cerebrum. Nitric oxide levels were no significantly different between all groups. Malonyldialdehyde levels increased in intoxication group (group 2) and decreased in treatment group (group 3). Activities of superoxide dismutase decreased in intoxication group (group 2) and increased in treatment group (group 3). Activities of catalase increased in intoxication group (group 2) and decreased in treatment group (group 3). Activities of glutathione peroxidase (GSH-Px) decreased in intoxication group (group 4) and increased in treatment group (group 5). CO poisoning caused significant damage, detected within the first 6 hours. Due to antioxidant enzymes, especially GSH-Px activity reaching the top level within 24th hours, significant oxidative damage was not observed. The protective effect against oxidative damage of magnesium sulfate has been identified within the first 6 hours.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Glutathione Peroxidase from bovine erythrocytes, lyophilized powder, ≥300 units/mg protein
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Catalase from Corynebacterium glutamicum, solution, deep brown, ≥500000 U/mL
Sigma-Aldrich
Catalase from bovine liver, powder, suitable for cell culture, 2,000-5,000 units/mg protein
Sigma-Aldrich
Catalase from Aspergillus niger, ammonium sulfate suspension, ≥4,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 40,000-60,000 units/mg protein (E1%/405)
Sigma-Aldrich
Superoxide Dismutase from human erythrocytes, essentially salt-free, lyophilized powder, ≥2,500 units/mg protein
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, 2,000-5,000 units/mg protein
Sigma-Aldrich
Catalase from human erythrocytes, ≥90% (SDS-PAGE), buffered aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, ≥10,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 10,000-40,000 units/mg protein
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, reagent grade, ≥97%
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
Superoxide Dismutase from bovine liver, lyophilized powder, ≥1500 units/mg protein
Sigma-Aldrich
Superoxide Dismutase from bovine liver, ammonium sulfate suspension, 2,000-6,000 units/mg protein (biuret)
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioReagent, for molecular biology, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Superoxide Dismutase from bovine erythrocytes, BioUltra, lyophilized powder, ≥4,500 units/mg protein, ≥97% (SDS-PAGE)
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Superoxide Dismutase bovine, recombinant, expressed in E. coli, lyophilized powder, ≥2500 units/mg protein, ≥90% (SDS-PAGE)
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioUltra, ≥99.5% (KT)
Sigma-Aldrich
Superoxide Dismutase from bovine erythrocytes, lyophilized powder, ≥3,000 units/mg protein
Sigma-Aldrich
Superoxide Dismutase I human, recombinant, expressed in E. coli
Sigma-Aldrich
Superoxide Dismutase from bovine erythrocytes, BioReagent, ≥3,000 units/mg protein, suitable for cell culture, lyophilized powder
Sigma-Aldrich
Magnesium sulfate heptahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Magnesium sulfate heptahydrate, 99.5-100.5% (calc. to the dried substance), meets analytical specification of Ph. Eur., BP,USP, FCC