  • Search Results
  • Myocardial toxicity of acute promyelocytic leukaemia drug-arsenic trioxide.
Myocardial toxicity of acute promyelocytic leukaemia drug-arsenic trioxide.
European review for medical and pharmacological sciences (2013-04-26)
V V Mathews, M V S Paul, M Abhilash, A Manju, S Abhilash, R H Nair
ABSTRACT
Arsenic trioxide (As2O3) is an environmental toxicant as well as an effective anti cancer agent against many types of cancers. It is a promising drug for patients with relapsed acute promyelocytic leukaemia (APL), but its clinical efficacy is burdened by the serious cardiac toxicities. The present study was designed to investigate the toxic mechanism of arsenic in cardiac tissue at its clinically relevant concentrations. Experimental rats were administered with As2O3 2, 4 and 8 mg/kg body weight, orally for a period of 45 days. Cardiac toxicities were recorded by lipid peroxidation, activities of glutathione dependent antioxidant and antiperoxidative enzymes, cardiac arsenic accumulation and histopathological changes. In vivo studies revealed a significant rise in lipid peroxidation, decline in reduced glutathione, glutathione dependent antioxidant enzymes and antiperoxidative enzymes in the cardiac tissue of arsenic treated rats. The extent of free radical production was found increased with periodic rise in the arsenic concentration. The experimental group which received 8 mg/kg body weight of arsenic exhibited the highest deposition of arsenic in cardiac tissue. Light microscopic examination of cardiac tissues in arsenic treated rats has showed increased structural abnormalities like myocardial fibre swelling, capillary congestion and micro-haemorrhages. The study concludes that the mechanism of arsenic induced cardiac toxicity is associated with the accumulation of arsenic in tissue and the extent of free radical production.
Product Description
Supelco
Arsenic(III) oxide, certified reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Superoxide Dismutase from bovine liver, ammonium sulfate suspension, 2,000-6,000 units/mg protein (biuret)
Sigma-Aldrich
Glutathione Reductase human, buffered aqueous solution, ≥10 units/mg protein, recombinant, expressed in E. coli
Sigma-Aldrich
Superoxide Dismutase from bovine erythrocytes, ≥3,000 units/mg protein, BioReagent, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Superoxide Dismutase from bovine liver, lyophilized powder, ≥1500 units/mg protein
Sigma-Aldrich
Superoxide Dismutase from bovine erythrocytes, BioUltra, lyophilized powder, ≥4,500 units/mg protein, ≥97% (SDS-PAGE)
Sigma-Aldrich
Superoxide Dismutase bovine, recombinant, expressed in E. coli, lyophilized powder, ≥2500 units/mg protein, ≥90% (SDS-PAGE)
Sigma-Aldrich
Arsenic(III) oxide, 99.995% trace metals basis
Sigma-Aldrich
Superoxide Dismutase I human, recombinant, expressed in E. coli
Sigma-Aldrich
Superoxide Dismutase from bovine erythrocytes, lyophilized powder, ≥3,000 units/mg protein
Sigma-Aldrich
Arsenic(III) oxide, ACS reagent (primary standard)
Sigma-Aldrich
Arsenic(III) oxide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Catalase from Aspergillus niger, ammonium sulfate suspension, ≥4,000 units/mg protein
Sigma-Aldrich
Catalase from human erythrocytes, ≥90% (SDS-PAGE), buffered aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, 2,000-5,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 10,000-40,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, powder, suitable for cell culture, 2,000-5,000 units/mg protein
Sigma-Aldrich
Glutathione Peroxidase from bovine erythrocytes, lyophilized powder, ≥300 units/mg protein
Sigma-Aldrich
Glutathione S-Transferase from equine liver, lyophilized powder, ≥25 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, ≥10,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 40,000-60,000 units/mg protein (E1%/405)
Sigma-Aldrich
Catalase from Corynebacterium glutamicum, solution, deep brown, ≥500000 U/mL
Sigma-Aldrich
Superoxide Dismutase from human erythrocytes, essentially salt-free, lyophilized powder, ≥2,500 units/mg protein