• Início
  • Resultados da busca
  • Detection of Staphylococcus aureus with a fluorescence in situ hybridization that does not require lysostaphin.
Detection of Staphylococcus aureus with a fluorescence in situ hybridization that does not require lysostaphin.
Journal of clinical laboratory analysis (2011-03-26)
Thomas S Lawson, Russell E Connally, Jonathan R Iredell, Subramanyam Vemulpad, James A Piper
RESUMO
To detect with whole-cell fluorescence in situ hybridization (FISH), Staphylococcus aureus is typically permeabilized with lysozyme and lysostaphin. We tested whether it was feasible to detect S. aureus and differentiate it from Staphylococcus epidermidis with lysozyme-only permeabilization. We compared lysozyme permeabilization to S. aureus permeabilized with lysozyme in combination with lysostaphin. It was determined that S. aureus treated with agarose, methanol, and lysozyme could be detected with FISH. The 1  hr protocol is a useful alternative to conventional FISH.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, lyophilized powder, Protein 50-70 % by biuret, ≥500 units/mg protein
Sigma-Aldrich
Proteinase K from Tritirachium album, buffered aqueous glycerol solution, for molecular biology, ≥800 units/mL
Sigma-Aldrich
Lysostaphin from Staphylococcus simulans, recombinant, expressed in E. coli, lyophilized powder
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, BioUltra, ≥97% (SDS-PAGE), Protein 40-60 % by biuret, ≥2,000 units/mg protein
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, aseptically filled