• Início
  • Resultados da busca
  • Water-soluble extract of Pacific Krill prevents triglyceride accumulation in adipocytes by suppressing PPARγ and C/EBPα expression.
Water-soluble extract of Pacific Krill prevents triglyceride accumulation in adipocytes by suppressing PPARγ and C/EBPα expression.
PloS one (2011-07-16)
Hidetoshi Yamada, Tomohiro Ueda, Akira Yano
RESUMO
Pacific Krill (Euphausia pacifica) are small, red crustaceans, similar to shrimp, that flourish in the North Pacific and are eaten in Japan. We investigated the effect of a water-soluble extract of Pacific Krill on adipocytes and discovered that this extract suppressed triglyceride accumulation in adipocytes. Furthermore, the water-soluble extract of Pacific Krill suppressed the expression of two master regulators of adipocyte differentiation, peroxisome proliferator-activated receptor gamma (PPARγ) and CCAAT enhancer binding protein alpha (C/EBPα). C/EBPβ promotes PPARγ and C/EBPα expression, but the water-soluble extract of Pacific Krill did not inhibit the expression of C/EBPβ or C/EBPβ-mediated transcriptional activation. The Pacific Krill extract was more effective than a PPARγ antagonist in suppressing PPARγ and C/EBPα expression. These results indicated that the water-soluble extract of Pacific Krill was not simply a PPARγ antagonist, but that it prevented triglyceride accumulation in adipocytes by suppression of PPARγ and C/EBPα via a pathway that is independent of C/EBPβ.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Antibiotic Antimycotic Solution (100×), Stabilized, with 10,000 units penicillin, 10 mg streptomycin and 25 μg amphotericin B per mL, 0.1 μm filtered, BioReagent, suitable for cell culture