Crystallographic coincidence of two bridging species in a dinuclear Co ethynyl-benzene complex.
Acta crystallographica. Section E, Structure reports online (2011-08-13)
Wesley A Hoffert, Matthew P Shores
RESUMO
In the title compound, trans,trans-[μ-(m-phenyl-ene)bis-(ethyne-1,2-di-yl)]bis-[chlorido(1,4,8,11-tetra-aza-cyclo-tetra-deca-ne)cobalt(III)]-trans,trans-[μ-(5-bromo-m-phenyl-ene)bis-(ethyne-1,2-di-yl)]bis-[chlorido(1,4,8,11-tetra-aza-cyclo-tetra-deca-ne)cobalt(III)]-tetra-phenyl-borate-acetone (0.88/0.12/2/4), [Co(2)(C(12)H(4))Cl(2)(C(10)H(24)N(4))(2)](0.88)[Co(2)(C(10)H(3)Br)Cl(2)(C(10)H(24)N(4))(2)](0.12)(C(24)H(20)B)(2)·4C(3)H(6)O, with the exception of the acetyl-ene and bromine groups, all atomic postitions are the same in the two compounds and are modeled at full occupancy. The Co(III) ions are six-coordinate with acetyl-ide and chloride ligands bound to the axial sites and the N atoms from the cyclam rings coordinated at the equatorial positions. N-H⋯O and N-H⋯Cl hydrogen-bonding interactions help to consolidate the crystal packing.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%