Merck
Concentration dependence in kinetic arrest of the first-order magnetic transition in Ta doped HfFe2.
Journal of physics. Condensed matter : an Institute of Physics journal (2013-01-15)
R Rawat, P Chaddah, Pallab Bag, P D Babu, V Siruguri
RESUMO
Magnetic behavior of the pseudo-binary alloy Hf(1-x)Ta(x)Fe(2) has been studied, for which the zero-field ferromagnetic (FM) to antiferromagnetic (AFM) transition temperature is tuned near to T = 0 K. Our studies show that such composition lies around x = 0.230. Detailed magnetization studies on x = 0.225, 0.230 and 0.235 show thermomagnetic irreversibility at low temperature due to kinetic arrest of the first-order AFM-FM transition. All three compositions studied show a reentrant transition in the zero-field-cooled warming curve and non-monotonic variation of the upper critical field. The region in H-T space where these features of kinetic arrest manifest themselves increases with increasing Ta concentration.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Tantalum, foil, thickness 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, nanopowder, <25 nm particle size (BET), ≥99% trace metals basis
Sigma-Aldrich
Tantalum, powder, −325 mesh, 99.9% trace metals basis
Sigma-Aldrich
Tantalum, powder, 60-100 mesh, 99.9%
Sigma-Aldrich
Hafnium, turnings, crystal bar, 99.7% trace metals basis
Sigma-Aldrich
Tantalum, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, powder, 99.99% trace metals basis
Sigma-Aldrich
Tantalum, rod, diam. 6.35 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, wire, diam. 1.0 mm, ≥99.9% trace metals basis