  • Search Results
  • The effects of gender, age and region on macro- and micronutrient contents and fatty acid profiles in the muscles of roe deer and wild boar in Mecklenburg-Western Pomerania (Germany).
The effects of gender, age and region on macro- and micronutrient contents and fatty acid profiles in the muscles of roe deer and wild boar in Mecklenburg-Western Pomerania (Germany).
Meat science (2013-02-05)
D Dannenberger, G Nuernberg, K Nuernberg, E Hagemann
ABSTRACT
Samples of M. longissimus were collected from a total of 203 feral roe deer (n=118) and wild boar (n=85) in two regions of Mecklenburg-Western Pomerania (Germany). The muscle lipid saturated fatty acid proportions of roe deer and wild boar ranged between 33 and 49g/100g total fatty acids and 31 and 35g/100g total fatty acids, respectively. The total n-3 PUFA proportions in roe deer muscle varied between 8.0 and 14g/100g fatty acids, and in wild boar muscle between 2.6 and 6.0g/100g fatty acids. The major vitamin E homologue, α-tocopherol, was determined to be between 5.8 and 13.1mg/kg in roe deer muscles. Lower levels between 1.2 and 4.7mg/kg were measured in wild boar muscles. The iron and zinc concentrations in roe deer and wild boar muscle ranged from 26.3 to 33.9mg/kg and from 17.0 to 21.7mg/kg, and from 13.6 to 39.3mg/kg and 18.1 to 31.9mg/kg, respectively.
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Sigma-Aldrich
DL-α-Tocopherol acetate, ≥96% (HPLC)
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
Sigma-Aldrich
DL-α-Tocopherol acetate, tested according to Ph. Eur.
Supelco
Tocopheryl Acetate, a, Pharmaceutical Secondary Standard; Certified Reference Material
α-Tocopherol acetate, European Pharmacopoeia (EP) Reference Standard