Merck
Rhenium mixed-ligand complexes with S,N,S-tridentate thiosemicarbazone/thiosemicarbazide ligands.
Dalton transactions (Cambridge, England : 2003) (2013-02-13)
Pedro I da S Maia, Hung Huy Nguyen, Adelheid Hagenbach, Silke Bergemann, Ronald Gust, Victor M Deflon, Ulrich Abram
RESUMO
Rhenium(V) complexes containing tridentate thiosemicarbazones/thiosemicarbazides (H2L1) derived from N-[N′,N′-dialkylamino(thiocarbonyl)]benzimidoyl chlorides with 4,4-dialkylthiosemicarbazides have been synthesized by ligand-exchange reactions starting from [ReOCl(L1)]. The chlorido ligand of [ReOCl(L1)] (4) is readily replaced and reactions with ammonium thiocyanate or potassium cyanide give [ReO(NCS)(L1)] (6) and [ReO(CN)(L1)] (7), respectively. The reaction of (NBu4)[ReOCl4] with H2L1 and two equivalents of ammonium thiocyanate, however, gives in a one-pot reaction [ReO(NCS)2(HL1)] (8), in which the pro-ligand H2L1 is only singly deprotonated. An oxo-bridged, dimeric nitridorhenium(V) compound of the composition [{ReN(HL1)}2O] (11) is obtained from a reaction of (NBu4)[ReOCl4], H2L1 and sodium azide. The six-coordinate complexes [ReO(L1)(Ph2btu)] (12), where HPh2btu is N,N-diphenyl-N′-benzoylthiourea, can be obtained by treatment of [ReOCl(L1)] with HPh2btu in the presence of NEt3. Studies of the antiproliferative effects of the [ReOX(L1)] system (X = Cl−, NCS− or CN−) on breast cancer cells show that the lability of a monodentate ligand seems to play a key role in the cytotoxic activity of the metal complexes, while the substitution of this ligand by the chelating ligand Ph2btu− completely terminates the cytotoxicity.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Rhenium, powder, −100 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Rhenium, foil, thickness 1.0 mm, 99.98% trace metals basis
Sigma-Aldrich
Rhenium, powder, 99.995% trace metals basis
Sigma-Aldrich
Rhenium, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Thiosemicarbazide, 99%
Sigma-Aldrich
Thiosemicarbazide, puriss. p.a., 98%