• Início
  • Resultados da busca
  • Site-specific N-glycosylation of caprine lysostaphin restricts its bacteriolytic activity toward Staphylococcus aureus.
Site-specific N-glycosylation of caprine lysostaphin restricts its bacteriolytic activity toward Staphylococcus aureus.
Animal biotechnology (2013-03-29)
Ching-Ying Huang, Jih-Tay Hsu, Pei-Hsuan Chung, Winston Teng-Kuei Cheng, Yan-Nian Jiang, Yu-Ten Ju
RESUMO
Lysostaphin (LYS) is an anti-staphylococcal prokaryotic polypeptide that has been used to avoid Staphylococcus aureus mastitis through transgenic or viral vector approaches exogenously expressed in dairy animals. However, glycosylation of lysostaphin expressed in mammalian cells results in a loss of bioactivity. Until now, the mechanism of site-specific glycosylation of lysostaphin causing this loss of bioactivity remains unknown. An immortalized caprine mammary epithelial cell line (CMEC-08-D) was used to study recombinant lysostaphin fused with goat β-casein, goat lactoferrin (LF) or prokaryotic signal peptides. These constructs were separately ectopically expressed in CMEC-08-D. Results of site-directed mutagenesis show that Asn(125) but not Asn(232) is the exact glycosylation site of lysostaphin expressed in CMEC-08-D. In addition, the effect of glycosylation of lysostaphin on its staphylolytic activity was identified through bacterial plate assay. The data indicated that wild type and mutated N232Q-lysostaphin (Asn(232) to Gln(232) substitution) lacked staphylolytic activity. In contrast, mutated N125Q (Asn(125) to Gln(125) substitution) and N125Q/N232Q-lysostaphin possessed staphylolytic activity. On the other hand, all mutated lysostaphin showed no change in binding ability to S. aureus. This reveals that N-glycosylation at Asn(125) of lysostaphin expressed in a eukaryotic system greatly decreases lysostaphin bacteriolytic activity but does not affect its binding ability to S. aureus.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, lyophilized powder, Protein 50-70 % by biuret, ≥500 units/mg protein
Sigma-Aldrich
Lysostaphin from Staphylococcus simulans, recombinant, expressed in E. coli, lyophilized powder
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, BioUltra, ≥97% (SDS-PAGE), Protein 40-60 % by biuret, ≥2,000 units/mg protein
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, free of DNA contaminants, suitable for Microbiome research, lyophilized powder, ≥500 units/mg protein
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, aseptically filled