  • Search Results
  • Synthesis of gold coated magnetic microparticles and their application for electrochemical glucose sensing by the enzymatically precipitated prussian blue.
Synthesis of gold coated magnetic microparticles and their application for electrochemical glucose sensing by the enzymatically precipitated prussian blue.
Journal of biomedical nanotechnology (2013-06-28)
Hye Young Jung, Jun Hui Park, Seongpil Hwang, Juhyoun Kwak
ABSTRACT
An enzyme stimulated deposition of prussian blue onto the gold-coated magnetic microparticles is described. We propose to synthesize the continuous outer gold layer on the magnetic particle for a gold working electrode and its superparamagnetic property. In-depth characterization of the gold shell formation was studied with scanning electron microscopy, energy-dispersive X-ray spectroscopy, cyclic voltammetry. The gold-coated magnetic microparticles offered adhesive layer for the immobilization of glucose oxidase catalyzing the generation of prussian blue in the presence of glucose. The assembled prussian blue on the gold shell surfaces was detected with electrochemical measurements depending on the glucose concentration. With accomplishing the linear response range from 0.2 mM to 10 mM of glucose, this approach successfully proposed the applicability of the magnetic core-gold shell structures to the electrochemical bioassay area.
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Supelco
D-(+)-Glucose solution, 1 mg/mL in 0.1% benzoic acid, standard for enzymatic assay kits GAGO20, GAHK20, STA20, analytical standard
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Iron(III) ferrocyanide
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C