  • Search Results
  • [Production of L-lactic acid from pentose by a genetically engineered Escherichia coli].
[Production of L-lactic acid from pentose by a genetically engineered Escherichia coli].
Wei sheng wu xue bao = Acta microbiologica Sinica (2013-07-19)
Jinfang Zhao, Liyuan Xu, Yongze Wang, Xiao Zhao, Jinhua Wang
ABSTRACT
In this study, we constructed a recombinant Escherichia coli strain for the production of high-purity L-lactic acid, using a homoethanol fermenting mutant E. coli SZ470 (deltafrdBC deltaldhA deltaackA deltafocA-pflB deltapdhR: :pflBp6-pflBrbs-aceEF-lpd) as the starting strain. By using homologous recombination, we deleted the adhE gene from SZ470 to obtain a mutant Escherichia coli JH01, which could not grow under anaerobic conditions. Then we cloned the L-lactate dehydrogenase gene (ldhL) of Pediococcus acidilactici and inserted it into the chromosome of JH01 via electroporation to obtain a recombinant strain Escherichia coli JH12. We evaluated the L-lactic acid production of the recombinant strain in a 15 L fermenter. In 10 L LB medium supplemented with 6% glucose, JH12 maintained maximal cell growth and an efficient L-lactic acid production rate for 36 h. Glucose consumption rate achieved was 1.46 g/(L x h) and L-lactic acid production rate was 1.14 g/(L x h). The results also show that 41.13 g/L lactic acid was produced, achieving a purity of 95.69% (based on total fermentation products). Xylose consumption rate was 0.88 g/(L x h) and L-lactic acid production rate was 0.60 g/(L x h). The production of lactic acid was 34.73 g/L, achieving a purity of 98%. There were no succinic acid and formic acid detected and only little amount of acetic acid generated during the fermentation. We constructed a homolactic acid fermentation strain E. coli JH12, which could efficiently convert glucose and xylose into high-purity L-lactic acid. JH12 could have great potential in industrial fermentation for L-lactic acid production.
Product Description
Supelco
Lactic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Lactic acid, natural, ≥85%
Sigma-Aldrich
Lactic acid, 85%, FCC
Sigma-Aldrich
Lactic acid, meets USP testing specifications
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
DL-Lactic acid, ~90% (T)
Sigma-Aldrich
Lactic acid solution, ACS reagent, ≥85%
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
DL-Lactic acid, 85 % (w/w), syrup
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Supelco
D-(+)-Glucose solution, 1 mg/mL in 0.1% benzoic acid, standard for enzymatic assay kits GAGO20, GAHK20, STA20, analytical standard
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material