• Início
  • Resultados da busca
  • Perovskite phase formation of monosized lead zirconate (PbZrO3) nanoparticles prepared by the sono-assisted co-precipitation method.
Perovskite phase formation of monosized lead zirconate (PbZrO3) nanoparticles prepared by the sono-assisted co-precipitation method.
Journal of nanoscience and nanotechnology (2013-07-19)
Piyanut Charoonsuk, Naratip Vittayakorn
RESUMO
The chemical reaction and phase evolution of perovskite lead zirconate (PbZrO3; PZ) nanoparticles, synthesized by the sono-assisted co-precipitation method, have been investigated. The nanopowders were characterized using the X-ray diffraction technique (XRD), Fourier transform infrared (FT-IR) spectroscopy, Raman spectroscopy, transmission electron microscope (TEM) and differential scanning calorimetry (DSC). The perovskite phase, PbZrO3, begins to form at 600 degrees C and was completed at 900 degrees C. During the reaction of PbZrO3, only the tetragonal zirconia (t-ZrO2) phase was formed as an intermediate phase with low temperature range. Only Raman spectroscopy can identify the intermediate tetragonal zirconia (t-ZrO2) phase in PbZrO3 powders during calcinations process. The change in amount of the t-ZrO2 phase in PbZrO3 powders was estimated from Raman spectra as a function of the calcination temperature. Observations by transmission electron microscopy revealed that the PbZrO3 powders have a uniform spherical shape with nanosized particles. The average size of the particles is about 10.60 +/- 2 nm with narrow size distribution.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Zirconium(IV) oxide, powder, 5 μm, 99% trace metals basis
Sigma-Aldrich
Zirconium(IV) oxide, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Zirconium, powder, −100 mesh
Sigma-Aldrich
Lead, powder, −100 mesh, 99.95% trace metals basis
Sigma-Aldrich
Lead, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Calcium titanate, nanopowder, <100 nm particle size (BET), 99% trace metals basis
Sigma-Aldrich
Zirconium(IV) oxide, nanoparticles, dispersion, <100 nm particle size (BET), 5 wt. % in H2O
Sigma-Aldrich
Zirconium, sponge, ≥99% trace metals basis
Sigma-Aldrich
Lead, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Zirconium, rod, diam. 6.35 mm, ≥99% trace metals basis
Sigma-Aldrich
Lead, shot, <2 mm, 99.9% trace metals basis
Sigma-Aldrich
Lead, shot, 1-3 mm, 99.995% trace metals basis
Sigma-Aldrich
Zirconium(IV) oxide, nanoparticles, dispersion, <100 nm particle size (BET), 10 wt. % in H2O
Supelco
Lead (impurities), manufactured by MBH Analytical Ltd (83X PR8)