  • Search Results
  • Superior photoluminescence and photocatalytic activity of CdS (core)-SiO2 (shell) nanostructures obtained by CdS photoetching and Au deposition.
Superior photoluminescence and photocatalytic activity of CdS (core)-SiO2 (shell) nanostructures obtained by CdS photoetching and Au deposition.
Journal of nanoscience and nanotechnology (2013-08-02)
Nidhi Gupta, Bonamali Pal
ABSTRACT
Core-shell morphology of silica (SiO2) coated CdS nanocomposites (SiO2@CdS) of different shapes have been made for better stability, luminescence and photochemical activity of CdS nanoparticles. A thin layer (thickness 1-1.4 nm) of SiO2 shell is deposited over CdS nanorods (CdS-NR) of aspect ratio = 21 and CdS nanospheres (CdS-NS) of size 6-8 nm by alkyl silane agents. Synthesized nanostructures were characterized by diffuse reflectance spectra, HR-TEM, BET surface measurement, LB surface film, and absorption and photoluminescence analysis. Photoetching (PE) of CdS core led to blue shift of the absorbance onset of SiO2@CdS-NR along with the appearance of an exciton band at 485 nm due to the quantum confinement effect. Photodissolution of CdS core shifts the band gap energy from initial 2.4 to 2.6 eV for CdS-NR and 2.5 to 2.67 eV for CdS-NS. TEM images reveal the increase in aspect ratio of NR from 21 to 31 and decrease in the spherical core to 2.5 nm from 6-8 nm after PE. Photoetched SiO2@CdS-NC displayed highly intense fluorescence emission (SiO2@CdS-NS > SiO2@CdS-NR) than unetched SiO2@CdS-NC at 488 nm corresponding to band edge position. The Au (0.5 wt.%) deposition onto photoetched SiO2@CdS-NR(PE) composites highly enhanced the fluorescence intensity in comparison to 1 wt.% of Au and Ag loading. SiO2@CdS-NC(PE) displayed improved photocatalytic activity during benzaldehyde photooxidation under UV (125 W, Hg-arc, 10.4 mW/cm2) irradiation. Silica coating onto CdS particles improves the photostability and photoactivity of CdS upon long UV irradiation.
Sigma-Aldrich
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Gold, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Gold, foil, thickness 0.05 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 1.5 mm, 99.999% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Silica, nanopowder, spec. surface area 175-225 m2/g (BET), 99.8% trace metals basis
Sigma-Aldrich
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
Sigma-Aldrich
Gold, wire, diam. 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 2.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, powder, <45 μm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Silica
Sigma-Aldrich
Silicon dioxide, alumina doped, nanoparticles, dispersion, <50 nm particle size, 20 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Gold, rod, diam. 3.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 1.0 mm, 99.997% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, beads, 1-6 mm, 99.999% trace metals basis
Sigma-Aldrich
Gold, powder, <10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silica, mesostructured, MSU-F (cellular foam)
Sigma-Aldrich
Gold coated microscope slide, layer thickness 100 Å, 99.999% (Au)
Sigma-Aldrich
Silica, mesostructured, MCM-41 type (hexagonal)
Sigma-Aldrich
Gold coated microscope slide, layer thickness 1000 Å, 99.999% (Au)
Sigma-Aldrich
Silicon dioxide, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
Sigma-Aldrich
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis