• Início
  • Resultados da busca
  • Preparation and photocatalytical performance of TiO2:SiO2 nanocomposites produced by the polymeric precursors method.
Preparation and photocatalytical performance of TiO2:SiO2 nanocomposites produced by the polymeric precursors method.
Journal of nanoscience and nanotechnology (2013-08-02)
Margaret Dawson, Gabriela Byzynski Soares, Caue Ribeiro
RESUMO
Anatase TiO2 is a promising photocatalyst due to its chemical stability, non-toxic characteristics, notable UV light absorption as well as photo-corrosion resistance and oxidative properties. Surface area and TiO2 dispersion quality are important factors that affect photoactivity of TiO2:SiO2 nanocomposites. In order to improve these factors, TiO2 nanoparticles were immobilized on mesoporous silica substrate through the polymeric precursors method, obtaining the nanocomposites in a simple routine. The TiO2 resin was synthesized by the polymeric precursors method and different resin thickness (0.5; 1.0; 2.0; 3.5; 5.0 nm) on silica were synthesized by calcination during 4 hours at 450 degrees C in pH 1.5. The selected pH for immobilization ensured adhesion of TiO2 nanoparticles onto the silica substrate surface. X-Ray Diffraction patterns indicate that all samples were predominantly anatase phase and immobilization improved surface area. Ametryn kinetic evaluation presents better results for SAM 3.5 and SAM 0.5. The results show that difference in TiO2 loading, surface area and crystallinity of samples are factors that influence photocatalytic efficiency.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Titanium(IV) oxide, nanopowder, 21 nm primary particle size (TEM), ≥99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, nanopowder, <25 nm particle size, 99.7% trace metals basis
Sigma-Aldrich
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, 99.8% trace metals basis
Sigma-Aldrich
LUDOX® AS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
Sigma-Aldrich
LUDOX® TM-50 colloidal silica, 50 wt. % suspension in H2O
Sigma-Aldrich
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, nanopowder, <100 nm particle size, 99.5% trace metals basis
Sigma-Aldrich
Silicon dioxide, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
Sigma-Aldrich
Silica, mesostructured, MCM-41 type (hexagonal)
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Silica, nanopowder, spec. surface area 175-225 m2/g (BET), 99.8% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5%
Sigma-Aldrich
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Titanium, foil, thickness 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
LUDOX® TMA colloidal silica, 34 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
Titanium, powder, <45 μm avg. part. size, 99.98% trace metals basis
Sigma-Aldrich
LUDOX® AS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Titanium, powder, −100 mesh, 99.7% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Sigma-Aldrich
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Titanium(IV) oxide, rutile, 99.995% trace metals basis
Sigma-Aldrich
Silica