• Início
  • Resultados da busca
  • Organic solar cells using a ZnO/Cu/ZnO anode deposited by ion beam sputtering at room temperature for flexible devices.
Organic solar cells using a ZnO/Cu/ZnO anode deposited by ion beam sputtering at room temperature for flexible devices.
Journal of nanoscience and nanotechnology (2013-08-02)
Ahmad El Hajj, Bruno Lucas, Anthony Barbot, Rémi Antony, Bernard Ratier, Matt Aldissi
RESUMO
The development of indium-free transparent conductive oxides (TCOs) on polymer substrates for flexible devices requires deposition at low temperatures and a limited thermal treatment. In this paper, we investigated the optical and electrical properties of ZnO/Cu/ZnO multi-layer electrodes obtained by ion beam sputtering at room temperature for flexible optoelectronic devices. This multilayer structure has the advantage of adjusting the layer thickness to favor antireflection and surface plasmon resonance of the metallic layer. We found that the optimal electrode is made up of a 10 nm-thick Cu layer between two 40 nm-thick ZnO layers, which results in a sheet resistance of 12 omega/(see symbol), a high transmittance of 85% in the visible range, and the highest figure of merit of 5.4 x 10(-3) (see symbol)/omega. A P3HT:PCBM-based solar cell showed a power conversion efficiency (PCE) of 2.26% using the optimized ZnO (40 nm)/Cu (10 nm)/ZnO (40 nm) anode.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Zinc oxide, nanopowder, <100 nm particle size
Sigma-Aldrich
Zinc oxide, nanopowder, <50 nm particle size (BET), >97%
Sigma-Aldrich
Zinc oxide, ReagentPlus®, powder, <5 μm particle size, 99.9%
Sigma-Aldrich
Copper, powder, <425 μm, 99.5% trace metals basis
Sigma-Aldrich
Zinc oxide, puriss. p.a., ACS reagent, ≥99.0% (KT)
Sigma-Aldrich
Zinc oxide, dispersion, nanoparticles, <100 nm particle size (TEM), ≤40 nm avg. part. size (APS), 20 wt. % in H2O
Sigma-Aldrich
Copper, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, powder, 99.999% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Zinc oxide, 99.99% trace metals basis
Sigma-Aldrich
Copper, ACS reagent, granular, 10-40 mesh, ≥99.90%
Sigma-Aldrich
Copper, nanopowder, 60-80 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
Copper, nanopowder, 40-60 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
Copper, powder (spheroidal), 10-25 μm, 98%
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, powder (dendritic), <45 μm, 99.7% trace metals basis
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
Zinc oxide, dispersion, nanoparticles, 40 wt. % in ethanol, <130 nm particle size
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc oxide, 99.999% trace metals basis
Sigma-Aldrich
Copper, bars (random sizes), ≥99.999% trace metals basis
Sigma-Aldrich
Copper, spherical shot, −20-+30 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Zinc oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. for dried substance)
Sigma-Aldrich
Copper, rod, diam. 11 mm, 99.9999% trace metals basis
Supelco
Zinc oxide, analytical standard
Sigma-Aldrich
Copper, shot, −3-+14 mesh, 99%