• Início
  • Resultados da busca
  • Synthesis and structural investigations of gel metal oxide composites WO₃-ZrO₂, WO₃-TiO₂, WO₃-ZrO₂-SiO₂, and their evaluation as materials for the preparation of ¹⁸⁸W/¹⁸⁸Re generator.
Synthesis and structural investigations of gel metal oxide composites WO₃-ZrO₂, WO₃-TiO₂, WO₃-ZrO₂-SiO₂, and their evaluation as materials for the preparation of ¹⁸⁸W/¹⁸⁸Re generator.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2013-03-19)
Edward Iller, Danuta Wawszczak, Marcin Konior, Halina Polkowska-Motrenko, Jacek J Milczarek, Ludwik Górski
RESUMO
The gel composites of WO3-ZrO2, WO3-TiO2, WO3-ZrO2-SiO2 at different molar ratios of oxides were synthesized. The elution profiles of the generator columns packed with gel samples activated at the Polish nuclear research reactor MARIA were studied. The best results of elution were found for WO3-ZrO2 and WO3-TiO2 both at 1:2M ratio annealed at 500°C and 650°C, respectively. Their structure determined with X-ray and neutron diffraction is completely amorphous with spherical grains of ∼23nm diameter. The efficiency of (188)W/(188)Re generators with WO3-ZrO2 gel of natural isotope composition as well as enriched in (186)W were investigated.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Tungsten, powder, 12 μm, 99.9% trace metals basis
Sigma-Aldrich
Tungsten, wire, diam. 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Rhenium, powder, 99.995% trace metals basis
Sigma-Aldrich
Tungsten, foil, thickness 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tungsten, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Tungsten, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tungsten, foil, thickness 0.127 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tungsten, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Rhenium, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Tungsten, foil, thickness 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Rhenium, powder, −100 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Rhenium, foil, thickness 1.0 mm, 99.98% trace metals basis