Using a geographic information system to improve childhood lead-screening efforts.
Preventing chronic disease (2013-06-15)
Robert Graff
RESUMO
The Idaho Division of Public Health conducted a pilot study to produce a lead-exposure-risk map to help local and state agencies better target childhood lead-screening efforts. Priority lead-screening areas, at the block group level, were created by using county tax assessor data and geographic information system software. A series of maps were produced, indicating childhood lead-screening prevalence in areas in which there was high potential for exposure to lead. These maps could enable development of more systematically targeted and cost-effective childhood lead-screening efforts.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Lead, powder, −100 mesh, 99.95% trace metals basis
Sigma-Aldrich
Lead, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Lead, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Lead, shot, <2 mm, 99.9% trace metals basis
Sigma-Aldrich
Lead, shot, 1-3 mm, 99.995% trace metals basis
Supelco
Lead (impurities), manufactured by MBH Analytical Ltd (83X PR8)