Evaluation of the fit of CAD/CAM abutments.
The International journal of prosthodontics (2013-07-10)
Adam Hamilton, Roy B Judge, Joseph E Palamara, Christopher Evans
RESUMO
This study aimed to compare the fit of computer-aided design/computerassisted manufacture (CAD/CAM) abutments provided by a single system with proprietary prefabricated abutments on various implant systems. Titanium CAD/CAM abutments were compared with prefabricated abutments on five different implant types. The samples were embedded in epoxy resin, sectioned longitudinally, and polished. Scanning electron microscopy was used to measure the gap between the implants and abutments at the connecting flanges and internal features. Independent t tests were used to compare data. A mean difference of 1.86 μm between the gold synOcta and CAD/CAM abutments on the Straumann Standard Plus implant was observed to be statistically significant (P = .002). Less than 0.4 μm of difference was found between the CAD/CAM and prefabricated abutments for the remaining implant types, and statistical significance was not observed. Mean differences of 34.4 μm (gold) and 44.7 μm (titanium) were observed between the CAD/ CAM and prefabricated abutments on the Straumann Standard Plus implants, which were statistically significant (P < .001). A mean difference of 15 μm was also observed between the CAD/CAM and prefabricated abutment on the NobelReplace implant, which was statistically significant (P = .026). All other groups had less that 4 μm of difference, and statistical significance was not observed. The CAD/CAM abutments appeared to have a comparable fit with prefabricated abutments for most of the systems evaluated. Design differences between the abutment connections for both Straumann implants were observed that affected the fit of internal components of the implant-abutment connections.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
Silicon carbide, −400 mesh particle size, ≥97.5%
Sigma-Aldrich
LUDOX® TM-50 colloidal silica, 50 wt. % suspension in H2O
Sigma-Aldrich
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
Sigma-Aldrich
Silicon dioxide, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
Sigma-Aldrich
Silica, mesostructured, MCM-41 type (hexagonal)
Sigma-Aldrich
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silica, nanopowder, spec. surface area 175-225 m2/g (BET), 99.8% trace metals basis
Sigma-Aldrich
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silicon carbide, -200 mesh particle size
Sigma-Aldrich
LUDOX® TMA colloidal silica, 34 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silicon carbide, nanopowder, <100 nm particle size
Sigma-Aldrich
Diamond, nanopowder, <10 nm particle size (TEM), ≥97% trace metals basis
Sigma-Aldrich
Silica
Sigma-Aldrich
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silica, mesostructured, MSU-F (cellular foam)
Sigma-Aldrich
LUDOX® CL-X colloidal silica, 45 wt. % suspension in H2O
Sigma-Aldrich
Diamond, nanopowder, <10 nm particle size (TEM), ≥95% trace metals basis
Sigma-Aldrich
Silicon dioxide, alumina doped, nanoparticles, dispersion, <50 nm particle size, 20 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Diamond, synthetic monocrystalline powder, ≤1 μm
Sigma-Aldrich
Diamond, synthetic polycrystalline powder, ~1 μm
Sigma-Aldrich
Diamond, monocrystalline powder, ~1 μm