  • Search Results
  • Relapsing visceral leishmaniasis in a HIV-1 infected patient with advanced disease.
Relapsing visceral leishmaniasis in a HIV-1 infected patient with advanced disease.
Acta clinica Belgica (2013-08-24)
D Standaert, F Laurent, S Jonckheere, J M Scheiff, B Vandercam, J C Yombi
ABSTRACT
Leishmaniasis, an intracellular protozoal infection in which tissue macrophages are targeted, is transmitted by female sandfly bite and occurs in 98 countries. Visceral leishmaniasis (VL) is the clinical form of leishmaniasis most frequently associated with HIV, especially in Europe. Both diseases have a synergistic detrimental effect on the cellular immune response. Treatment of VL in patients with underlying HIV-infection is associated with lower cure rates, higher rates of drug toxicity, higher relapse rates and greater mortality than treatment of VL in immunocompetent patients. We report the case of a HIV-1 infected patient with advanced disease who presented VL with multiple relapses. This case highlights the difficulties of treating VL in patients with HIV co-infection.
Sigma-Aldrich
Amphotericin B from Streptomyces sp., ~80% (HPLC), powder
Sigma-Aldrich
Amphotericin B from Streptomyces sp., BioReagent, suitable for cell culture, ~80% (HPLC)
Supelco
Amphotericin B trihydrate, VETRANAL®, analytical standard
Sigma-Aldrich
Amphotericin B solution, 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture
Amphotericin B for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Amphotericin B solubilized, powder, γ-irradiated, BioXtra, suitable for cell culture