• Início
  • Resultados da busca
  • Influence of dentin pretreatment with titanium tetrafluoride and self-etching adhesive systems on microtensile bond strength.
Influence of dentin pretreatment with titanium tetrafluoride and self-etching adhesive systems on microtensile bond strength.
American journal of dentistry (2013-08-31)
Enrico Coser Bridi, Flávia Lucisano Botelho Amaral, Fabiana Mantovani Gomes França, Cecília Pedroso Turssi, Roberta Tarkany Basting
RESUMO
To evaluate the effect of dentin pretreatment with 2.5% titanium tetrafluoride (TiF4) on microtensile bond strength (microTBS) of one- or two-step self-etching adhesive systems. 24 human sound third molars were used. A flat dentin surface of each tooth was exposed. After planing, teeth were divided into groups so that dentin would be left untreated or treated with a 2.5% TiF4 solution for 1 minute. Specimens were then subdivided into two groups to receive one of the following adhesive systems: one-step self-etching Adper Easy One (ADP) or two-step self-etching adhesive Clearfil SE Bond (CLEAR). A block of composite measuring 5.0 mm high and 5.0 mm wide was made incrementally on the tooth. Specimens were taken to a metallographic cutter to fabricate sticks with a bond area of approximately 1 mm2. After 24 hours, specimens were submitted to microTBS testing and the failure mode was recorded by examining specimens under stereomicroscopy. Scanning electron microscope (SEM) photomicrographs were obtained of the tooth/restoration interface. Two-way ANOVA and Tukey's test demonstrated that pretreatment of dentin with a TiF4 solution did not affect the microTBS values of either of the adhesive systems (P = 0.675). CLEAR provided higher bond strength than ADP, regardless of whether dentin was or was not pretreated with the TiF4 solution. Failure mode showed mostly adhesive failures in all groups, except when only ADP was used, causing mostly cohesive fractures in resin.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, powder, <45 μm avg. part. size, 99.98% trace metals basis
Sigma-Aldrich
Titanium, powder, −100 mesh, 99.7% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, sponge, 3-19 mm, 99.5% trace metals basis
Sigma-Aldrich
Titanium, nanoparticles, dispersion, <100 nm particle size, in mineral oil, 98.5% trace metals basis
Sigma-Aldrich
Titanium, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.995% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.81 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, crystalline, 5-10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Sigma-Aldrich
Titanium, evaporation slug, diam. × L 6.3 mm × 6.3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 2.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, sputtering target, diam. × thickness 3.00 in. × 0.125 in., 99.995% trace metals basis
Sigma-Aldrich
Titanium, rod, diam. 6.35 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.1 mm, 99.99% trace metals basis
Titanium, IRMM®, certified reference material, 0.5 mm wire
Titanium, IRMM®, certified reference material, 0.1 mm foil
Titanium, IRMM®, certified reference material, 0.5 mm foil