  • Search Results
  • Reversal of corticosteroid-induced diabetes mellitus with supplemental chromium.
Reversal of corticosteroid-induced diabetes mellitus with supplemental chromium.
Diabetic medicine : a journal of the British Diabetic Association (1999-05-06)
A Ravina, L Slezak, N Mirsky, N A Bryden, R A Anderson
ABSTRACT
To determine if the stress of corticosteroid treatment increases chromium (Cr) losses and if corticosteroid-induced diabetes (steroid diabetes) can be reversed by supplemental chromium. The effects of corticosteroid treatment on chromium losses of 13 patients 2 days prior to steroid administration and the first 3 days following treatment were determined. Since steroid-induced diabetes was associated with increased chromium losses and insufficient dietary chromium is associated with glucose intolerance and diabetes, we treated three patients with steroid-induced diabetes with 600 microg per day of chromium as chromium picolinate. Urinary chromium losses following corticosteroid treatment increased from 155+/-28 ng/d before corticosteroid treatment to 244+/-33 ng/d in the first 3 days following treatment. Chromium supplementation of patients with steroid-induced diabetes resulted in decreases in fasting blood glucose values from greater than 13.9 mmol/l (250 mg/dl) to less than 8.3 mmol/l (150 mg/dl). Hypoglycaemic drugs were also reduced 50% in all patients when given supplemental chromium. These data demonstrate that corticosteroid treatment increases chromium losses and that steroid-induced diabetes can be reversed by chromium supplementation. Follow-up, double-blind studies are needed to confirm these observations.
Sigma-Aldrich
Chromium, powder, ≥99% trace metals basis, <45 μm
Sigma-Aldrich
Chromium, powder, 99.5%, −100 mesh
Sigma-Aldrich
Chromium, chips, thickness ~2 mm, 99.5%
Sigma-Aldrich
Chromium, chips, 99.995% trace metals basis
Sigma-Aldrich
Chromium, sputtering target, diam. × thickness 3.00 in. × 0.125 in., 99.95% trace metals basis