  • Search Results
  • Determination of chromium in cerebrospinal fluid using electrothermal atomisation atomic absorption spectrometry.
Determination of chromium in cerebrospinal fluid using electrothermal atomisation atomic absorption spectrometry.
Journal of trace elements in medicine and biology : organ of the Society for Minerals and Trace Elements (GMS) (2003-01-18)
María Victorina Aguilar, Carmen José Mateos, María Carmen Martínez Para
ABSTRACT
A rapid method to determine the chromium content of cerebrospinal fluid (CSF) samples using electrothermal atomization atomic absorption spectrometry (ETA-AAS) with deuterium-arc background correction is described. The chromium concentration in CSF was evaluated by the standard addition method. Sample dilution (1 + 1) with 0.25% (m/v) Triton X-100 and 4.5% (v/v) HNO3 gave the best combination of sensitivity, reproducibility, and low blank reading compared with dilution using other solvents. Within-batch reproducibility was 3.2% for 20 CSF samples, between-batch reproducibility was 4.7%. CSF samples from 43 healthy volunteers collected in a manner designed to avoid contamination yielded chromium concentrations of 14.6 +/- 6.3 ng mL(-1).
Sigma-Aldrich
Chromium, powder, ≥99% trace metals basis, <45 μm
Sigma-Aldrich
Chromium, powder, 99.5%, −100 mesh
Sigma-Aldrich
Chromium, chips, thickness ~2 mm, 99.5%
Sigma-Aldrich
Chromium, chips, 99.995% trace metals basis
Sigma-Aldrich
Chromium, sputtering target, diam. × thickness 3.00 in. × 0.125 in., 99.95% trace metals basis