Merck
  • Início
  • Resultados da busca
  • Influence of simulated weightlessness on the oral pharmacokinetics of acetaminophen as a gastric emptying probe in man: a plasma and a saliva study.
Influence of simulated weightlessness on the oral pharmacokinetics of acetaminophen as a gastric emptying probe in man: a plasma and a saliva study.
Journal of clinical pharmacology (2003-10-11)
Peggy Gandia, Marie-Pierre Bareille, Sylvie Saivin, Anne Pavy Le-Traon, Michel Lavit, Antonio Guell, Georges Houin
RESUMO
This study evaluated the effect of simulated weightlessness on gastric emptying, using acetaminophen as a probe and -6 degrees head-down bed rest to simulate zero gravity. Eighteen volunteers were given 1 g of acetaminophen orally before the bed rest and at days 1, 18, and 80. Cmax, tmax, AUC0- infinity, AUC0-t, and t1/2 were calculated for plasma and saliva. The plasma Cmax showed a significant increase (10.43 microg/mL [day 1] to 14.74 microg/mL [day 80]), while tmax significantly decreased (1.41 h [day 1] to 0.91 h [day 80]). Similar results were obtained with saliva, and there were significant increases in the AUCs. The good correlation between the plasma and saliva data suggests that saliva sampling can be valid for acetaminophen pharmacokinetics. The changes in Cmax and tmax indicated more rapid drug absorption, which could have been as a result of faster gastric emptying or an increased blood flow to the intestine.
MATERIAIS
Número do produto
Marca
Descrição do produto
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Paracetamol, European Pharmacopoeia (EP) Reference Standard