  • Search Results
  • Vitamin E status and its dietary determinants in Taiwanese--results of the Nutrition and Health Survey in Taiwan 1993-1996.
Vitamin E status and its dietary determinants in Taiwanese--results of the Nutrition and Health Survey in Taiwan 1993-1996.
European journal of nutrition (2004-04-15)
Mei-Jyh Kang, Yi-Chin Lin, Wen-Hui Yeh, Wen-Harn Pan
ABSTRACT
A large proportion of Taiwanese are considered to have inadequate vitamin E intake according to Taiwanese RDA. To evaluate the vitamin E status in Taiwan using biochemical indicators, and to examine the influences of dietary factors. The Nutrition and Health Survey in Taiwan 1993-1996 was conducted using a multi-stage sampling scheme. Data of 3614 subjects (1728 males, and 1886 females) aged 4 years and above were included in the current analysis. Females had higher levels of serum alpha-tocopherol than males. Serum level of alpha-tocopherol significantly increased with age and blood lipids (p < 0.001). The prevalence rate of deficiency, assessed by the ratio of serum alpha-tocopherol to cholesterol+triglyceride(TG) < 1.59 micromol/mmol, was 1.4 % for Taiwanese aged 4 and above. The prevalence was 1.02 % for adults. This biochemical profile was superior in women compared to men. The age-serum vitamin E status was U-shaped, being poorest in teenagers. Geographical variation in vitamin E/cholesterol+TG ratio was not apparent across regions. An association was observed between serum vitamin E status and frequency of vitamin E supplements, fresh fruits, and 100 % fruit juices. An association was also seen with dietary intakes of vitamin C and vitamin E assessed by 24-hour recall. The prevalence rate of vitamin E deficiency in Taiwan was low. An association was observed between serum vitamin E status and dietary vitamins E and C intakes either from foods or supplements.
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g