Merck
  • Início
  • Resultados da busca
  • Trace element reference values in tissues from inhabitants of the European community. I. A study of 46 elements in urine, blood and serum of Italian subjects.
Trace element reference values in tissues from inhabitants of the European community. I. A study of 46 elements in urine, blood and serum of Italian subjects.
The Science of the total environment (1990-06-01)
C Minoia, E Sabbioni, P Apostoli, R Pietra, L Pozzoli, M Gallorini, G Nicolaou, L Alessio, E Capodaglio
RESUMO
Neutron activation analysis-electrothermal atomic absorption spectroscopy (ETA-AAS) and inductively coupled plasma atomic emission spectrometry (ICP-AES) have been used for the determination of 46 elements in urine, 35 in blood and 26 in serum of unexposed Italian subjects living in the same region (Lombardy). The results allowed the proposal of reference values for various elements determined in more than 350 healthy subjects, these being Ag, Al, As, Be, Bi, Cd, Co, Cr, Cu, Hg, Mn, Ni, Pb, Sb, Se, Tl, V, Zn, in urine; Ag, As, Bi, Cd, Cr, Co, Cu, Hg, Pb, Se, Tl, Zn in blood; and Ag, Al, Be, Cd, Co, Cr, Cu, Hg, Mn, Ni, Pb, Se, Tl, V, Zn in serum (or plasma). For all other elements indicative values are suggested. In addition to the mean value and the "reference range", a "range of uncertainty" and an upper limit above which metabolic abnormalities could be expected have also been defined on the basis of simple statistical considerations.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Tantalum, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, powder, 99.99% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, rod, diam. 6.35 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, wire, diam. 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, nanopowder, <25 nm particle size (BET), ≥99% trace metals basis
Sigma-Aldrich
Tantalum, powder, −325 mesh, 99.9% trace metals basis
Sigma-Aldrich
Tantalum, powder, 60-100 mesh, 99.9%