Thrombosis of Björk-Shiley aortic valve prostheses.
Radiology (1980-10-01)
K Venkataraman, R F Beer, N P Mathews, J R Carl, E C Harrison, A F Turner, E J Finck
RESUMO
Though thromboembolic complications are reportedly minimal in patients with Björk-Shiley aortic valve prostheses, massive thrombosis remains a major and often fatal problem. The authors observed massive thrombosis in 3 patients, only one of whom survived. In all three instances, the disk of the prosthesis was relatively radiolucent. With valves manufactured after late 1975, which contain a tantalum foil hoop, cineradiography is the most rapid and accurate method of showing the disk. A simple method of calculating the valve opening angle from the cineradiograph is described. It is suggested that serial cineradiographs might be useful in detecting lesser degrees of thrombosis and partial immobilization of the disk.
MATERIAIS
Número do produto
Marca
Descrição do produto
Tantalum, foil, 10mm disks, thickness 0.05mm, annealed, 99.9%
Tantalum, foil, 10mm disks, thickness 0.075mm, annealed, 99.9%
Tantalum, foil, 10mm disks, thickness 0.100mm, annealed, 99.9%
Tantalum, foil, 10mm disks, thickness 0.125mm, annealed, 99.9%
Tantalum, foil, 10mm disks, thickness 0.15mm, annealed, 99.9%
Tantalum, foil, 10mm disks, thickness 0.20mm, annealed, 99.9%
Tantalum, foil, 10mm disks, thickness 0.25mm, annealed, 99.9%
Tantalum, foil, 10mm disks, thickness 0.25mm, annealed, 99.99%
Tantalum, foil, 10mm disks, thickness 0.5mm, annealed, 99.9%
Tantalum, foil, 10mm disks, thickness 0.75mm, annealed, 99.9%
Tantalum, foil, 15mm disks, thickness 0.006mm, 99.9%
Tantalum, foil, 15mm disks, thickness 0.0075mm, 99.9%
Tantalum, foil, 15mm disks, thickness 0.009mm, 99.9%
Tantalum, foil, 15mm disks, thickness 0.0125mm, 99.9%
Tantalum, foil, 15mm disks, thickness 0.015mm, 99.9%
Tantalum, foil, 15mm disks, thickness 0.01mm, 99.9%
Tantalum, foil, 15mm disks, thickness 0.025mm, as rolled, 99.9%
Tantalum, foil, 15mm disks, thickness 0.02mm, 99.9%
Tantalum, foil, 15mm disks, thickness 0.034mm, as rolled, 99.9%
Tantalum, foil, 15mm disks, thickness 0.03mm, as rolled, 99.9%
Tantalum, foil, 15mm disks, thickness 0.05mm, annealed, 99.9%
Tantalum, foil, 15mm disks, thickness 0.075mm, annealed, 99.9%
Tantalum, foil, 15mm disks, thickness 0.100mm, annealed, 99.9%
Tantalum, foil, 15mm disks, thickness 0.125mm, annealed, 99.9%
Tantalum, foil, 15mm disks, thickness 0.15mm, annealed, 99.9%
Tantalum, foil, 15mm disks, thickness 0.20mm, annealed, 99.9%
Tantalum, foil, 15mm disks, thickness 0.25mm, annealed, 99.9%
Tantalum, foil, 15mm disks, thickness 0.25mm, annealed, 99.99%
Tantalum, foil, 15mm disks, thickness 0.5mm, annealed, 99.9%
Tantalum, foil, 15mm disks, thickness 0.75mm, annealed, 99.9%
Redes sociais
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
Pesquisa. Desenvolvimento. Produção.
Somos um fornecedor global líder para o setor de ciências biológicas, com soluções e serviços para pesquisa, desenvolvimento e produção de biotecnologia, além de produção e desenvolvimento de terapias farmacológicas com medicamentos.
© 2021 Merck KGaA, Darmstadt, Alemanha e/ou suas filiais. Todos os direitos reservados.
A reprodução não autorizada de quaisquer materiais deste site é estritamente proibida.