Merck
Onyx embolization of a ruptured lenticulostriate artery aneurysm in a patient with moyamoya disease.
World neurosurgery (2012-04-10)
Nohra Chalouhi, Stavropoula Tjoumakaris, L Fernando Gonzalez, Aaron S Dumont, Qaisar Shah, David Gordon, Robert Rosenwasser, Pascal Jabbour
RESUMO
Lenticulostriate artery (LSA) aneurysms are uncommon. Most reported cases have been surgically treated. There are only three reports of LSA aneurysms treated with endovascular techniques. We report the case of a 49-year-old man who presented with a hemorrhage in the basal ganglia. An angiogram revealed a distal LSA aneurysm in the setting of moyamoya disease. The patient's aneurysm was successfully embolized by the use of Onyx 18. In this report, we present the first case of onyx embolization of a LSA aneurysm and review all the previous cases that were managed with endovascular techniques. Treatment of these lesions with either surgery or endovascular therapy is challenging and requires specialized expertise. Given the risks of surgery, we believe that LSA aneurysms are best treated by endovascular means, especially in the setting of moyamoya disease.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Tantalum, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, powder, 99.99% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, rod, diam. 6.35 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, wire, diam. 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, nanopowder, <25 nm particle size (BET), ≥99% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, powder, −325 mesh, 99.9% trace metals basis
Sigma-Aldrich
Tantalum, powder, 60-100 mesh, 99.9%