Merck
High survivorship with a titanium-encased alumina ceramic bearing for total hip arthroplasty.
Clinical orthopaedics and related research (2013-04-02)
James A D'Antonio, William N Capello, Marybeth Naughton
RESUMO
Although ceramic-on-ceramic bearings for total hip arthroplasty (THA) show promising results in terms of bearing-surface wear, fracture of the bearing, insertional chips, and squeaking remain a concern. Our primary objective of this report was to determine overall survivorship of a titanium-encased ceramic-on-ceramic bearing couple. Our secondary objectives were to evaluate for ceramic fracture, insertional chips, osteolysis, and device squeaking. Six surgeons at six institutions implanted 194 patients (209 hips) with an average age of 52 years with cementless hips and alumina ceramic bearings. One hundred thirty-seven patients (146 hips) have 10-year followup (70%). We determined Kaplan-Meier survivorship of the bearing surface and implant system and collected radiographic and clinical data to evaluate for osteolysis and squeaking. Survivorship using revision for any reason as the end point was 97% at 10 years and survivorship end point bearing surface failure or aseptic loosening of 99%. There was one ceramic insert fracture (0.5%), there were no insertional chips, there was no visible osteolysis on AP and lateral radiographs, and there was a 1% patient-self-reported incidence of squeaking at the last clinical followup. Six hips underwent revision (3.7%). Ceramic bearings for THA with a titanium-encased insert have high survivorship at 10 years followup and a fracture risk of 0.5%. We found at last followup on routine radiographs no evidence of osteolysis, and no patient has been revised for squeaking or has reported dissatisfaction with the clinical result because of noise. IV, therapeutic study. See the Guidelines for Authors for a complete description of levels of evidence.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Supelco
Aluminum oxide, for the determination of hydrocarbons
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Aluminum oxide, single crystal substrate, <0001>
Sigma-Aldrich
Aluminum oxide, activated, acidic, Brockmann I
Sigma-Aldrich
Aluminum oxide, activated, basic, Brockmann I
Sigma-Aldrich
Aluminum oxide, activated, neutral, Brockmann I
Sigma-Aldrich
Aluminum oxide, activated, acidic, Brockmann I, free-flowing, Redi-Dri
Sigma-Aldrich
Aluminum oxide, activated, neutral, Brockmann I, free-flowing, Redi-Dri
Sigma-Aldrich
Aluminum oxide, pellets, 3 mm
Sigma-Aldrich
Aluminum oxide, pore size 58 Å, ~150 mesh
Sigma-Aldrich
Aluminum oxide, Type WN-6, Neutral, Activity Grade Super I
Sigma-Aldrich
Aluminum oxide, powder, primarily α phase, ≤10 μm avg. part. size, 99.5% trace metals basis
Sigma-Aldrich
Aluminum oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, -325 mesh
Sigma-Aldrich
Aluminum oxide, calcined, powder, primarily α-phase, 100-325 mesh
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, 100-200 mesh
Sigma-Aldrich
Aluminum oxide, Corundum, α-phase, -100 mesh
Sigma-Aldrich
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O