  • Search Results
  • Neolignan Licarin A presents effect against Leishmania (Leishmania) major associated with immunomodulation in vitro.
Neolignan Licarin A presents effect against Leishmania (Leishmania) major associated with immunomodulation in vitro.
Experimental parasitology (2013-07-31)
Patrícia L N Néris, John P A Caldas, Yara K S Rodrigues, Francianne M Amorim, Jacqueline A Leite, Sandra Rodrigues-Mascarenhas, José M Barbosa-Filho, Luis C Rodrigues, Márcia R Oliveira
ABSTRACT
Leishmaniasis' treatment is based mostly on pentavalent antimonials or amphotericin B long-term administration, expensive drugs associated with severe side effects. Considering these aforementioned, the search for alternative effective and safe leishmaniasis treatments is a necessity. This work evaluated a neolignan, licarin A anti-leishmanial activity chemically synthesized by our study group. It was observed that licarin A effectively inhibited Leishmania (Leishmania) major promastigotes (IC₅₀ of 9.59 ± 0.94 μg/mL) growth, by inducing in these parasites genomic DNA fragmentation in a typical death pattern by apoptosis. Additionally, the neolignan proved to be even more active against intracellular amastigotes of the parasite (EC₅₀ of 4.71 ± 0.29 μg/mL), and significantly more effective than meglumine antimoniate (EC₅₀ of 216.2 ± 76.7 μg/mL) used as reference drug. The antiamastigote activity is associated with an immunomodulatory activity, since treatment with licarin A of the infected macrophages induced a decrease in the interleukin (IL)-6 and IL-10 production. This study demonstrates for the first time the antileishmanial activity of licarin A and suggests that the compound may be a promising in the development of a new leishmanicidal agent.
Sigma-Aldrich
Meglumine, meets USP testing specifications
Sigma-Aldrich
Amphotericin B solubilized, powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Amphotericin B from Streptomyces sp., ~80% (HPLC), powder
Sigma-Aldrich
Amphotericin B from Streptomyces sp., BioReagent, suitable for cell culture, ~80% (HPLC)
Sigma-Aldrich
N-Methyl-D-glucamine, 99.0-100.5% (titration)
Supelco
Amphotericin B trihydrate, VETRANAL®, analytical standard
Sigma-Aldrich
N-Methyl-D-glucamine, ReagentPlus®, ≥99.0% (T)
Sigma-Aldrich
Amphotericin B solution, 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture
Amphotericin B for peak identification, European Pharmacopoeia (EP) Reference Standard