Merck
  • Início
  • Resultados da busca
  • Enantioselective silyl protection of alcohols promoted by a combination of chiral and achiral Lewis basic catalysts.
Enantioselective silyl protection of alcohols promoted by a combination of chiral and achiral Lewis basic catalysts.
Nature chemistry (2013-08-24)
Nathan Manville, Hekla Alite, Fredrik Haeffner, Amir H Hoveyda, Marc L Snapper
RESUMO
Catalytic enantioselective monosilylations of diols and polyols furnish valuable alcohol-containing molecules in high enantiomeric purity. These transformations, however, require high catalyst loadings (20-30 mol%) and long reaction times (2-5 days). Here, we report that a counterintuitive strategy involving the use of an achiral co-catalyst structurally similar to the chiral catalyst provides an effective solution to this problem. A combination of seemingly competitive Lewis basic molecules can function in concert such that one serves as an achiral nucleophilic promoter and the other performs as a chiral Brønsted base. On the addition of 7.5-20 mol% of a commercially available N-heterocycle (5-ethylthiotetrazole), reactions typically proceed within one hour, and deliver the desired products in high yields and enantiomeric ratios. In some instances, there is no reaction in the absence of the achiral base, yet the presence of the achiral co-catalyst gives rise to facile formation of products in high enantiomeric purity.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Silicon, powder, −60 mesh, 99.998% trace metals basis
Sigma-Aldrich
Silicon, powder, −325 mesh, 99% trace metals basis
Sigma-Aldrich
Silicon, pieces, 99.95% trace metals basis
Sigma-Aldrich
Silicon, wafer, <111>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.3 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.999% trace metals basis
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), contains phosphorus as dopant, <111>, N-type, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <111>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, nanopowder, <100 nm particle size (TEM), ≥98% trace metals basis