• Início
  • Resultados da busca
  • Bradykinin-induced Ca2+ signaling in human subcutaneous fibroblasts involves ATP release via hemichannels leading to P2Y12 receptors activation.
Bradykinin-induced Ca2+ signaling in human subcutaneous fibroblasts involves ATP release via hemichannels leading to P2Y12 receptors activation.
Cell communication and signaling : CCS (2013-09-21)
Ana Rita Pinheiro, Diogo Paramos-de-Carvalho, Mariana Certal, Cristina Costa, Maria Teresa Magalhães-Cardoso, Fátima Ferreirinha, Maria Adelina Costa, Paulo Correia-de-Sá
RESUMO
Chronic musculoskeletal pain involves connective tissue remodeling triggered by inflammatory mediators, such as bradykinin. Fibroblast cells signaling involve changes in intracellular Ca2+ ([Ca2+]i). ATP has been related to connective tissue mechanotransduction, remodeling and chronic inflammatory pain, via P2 purinoceptors activation. Here, we investigated the involvement of ATP in bradykinin-induced Ca2+ signals in human subcutaneous fibroblasts. Bradykinin, via B2 receptors, caused an abrupt rise in [Ca2+]i to a peak that declined to a plateau, which concentration remained constant until washout. The plateau phase was absent in Ca2+-free medium; [Ca2+]i signal was substantially reduced after depleting intracellular Ca2+ stores with thapsigargin. Extracellular ATP inactivation with apyrase decreased the [Ca2+]i plateau. Human subcutaneous fibroblasts respond to bradykinin by releasing ATP via connexin and pannexin hemichannels, since blockade of connexins, with 2-octanol or carbenoxolone, and pannexin-1, with 10Panx, attenuated bradykinin-induced [Ca2+]i plateau, whereas inhibitors of vesicular exocytosis, such as brefeldin A and bafilomycin A1, were inactive. The kinetics of extracellular ATP catabolism favors ADP accumulation in human fibroblast cultures. Inhibition of ectonucleotidase activity and, thus, ADP formation from released ATP with POM-1 or by Mg2+ removal from media reduced bradykinin-induced [Ca2+]i plateau. Selective blockade of the ADP-sensitive P2Y12 receptor with AR-C66096 attenuated bradykinin [Ca2+]i plateau, whereas the P2Y1 and P2Y13 receptor antagonists, respectively MRS 2179 and MRS 2211, were inactive. Human fibroblasts exhibited immunoreactivity against connexin-43, pannexin-1 and P2Y12 receptor. Bradykinin induces ATP release from human subcutaneous fibroblasts via connexin and pannexin-1-containing hemichannels leading to [Ca2+]i mobilization through the cooperation of B2 and P2Y12 receptors.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Adenosine 5′-diphosphate, ≥95% (HPLC)
Sigma-Aldrich
1,3-Dimethyl-8-phenylxanthine, crystalline
Redes sociais
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
Pesquisa. Desenvolvimento. Produção.
Somos um fornecedor global líder para o setor de ciências biológicas, com soluções e serviços para pesquisa, desenvolvimento e produção de biotecnologia, além de produção e desenvolvimento de terapias farmacológicas com medicamentos.
© 2021 Merck KGaA, Darmstadt, Alemanha e/ou suas filiais. Todos os direitos reservados.
A reprodução não autorizada de quaisquer materiais deste site é estritamente proibida.