  • Search Results
  • Equilibrium between a cyclotrisilene and an isolable base adduct of a disilenyl silylene.
Equilibrium between a cyclotrisilene and an isolable base adduct of a disilenyl silylene.
Nature chemistry (2013-09-24)
Michael J Cowley, Volker Huch, Henry S Rzepa, David Scheschkewitz
ABSTRACT
In organic chemistry, compounds with adjacent alkene and carbene functionalities (vinyl carbenes) are studied widely as fleeting intermediates and in the coordination sphere of transition metals. Stable derivatives of vinyl carbenes remain elusive, including the corresponding heavier group 14 homologues. Here we report the isolation and full characterization of a base-stabilized silicon version of a vinyl carbene that features a silicon-silicon double bond as well as a silylene functionality, coordinated by an N-heterocyclic carbene (NHC). In solution, the intensely green disilenyl silylene adduct exists in equilibrium with the corresponding silicon analogue of a cyclopropene and free NHC, which was quantified by nuclear magnetic resonance spectroscopy and ultraviolet-visible spectroscopy. The reversibility of this process raises exciting possibilities for the preparation of extended conjugated π systems of silicon.
Sigma-Aldrich
Silicon, powder, −60 mesh, 99.998% trace metals basis
Sigma-Aldrich
Silicon, pieces, 99.95% trace metals basis
Sigma-Aldrich
Silicon, powder, −325 mesh, 99% trace metals basis
Sigma-Aldrich
Silicon, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), contains phosphorus as dopant, <111>, N-type, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <111>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer, <111>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.3 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, nanopowder, <100 nm particle size (TEM), ≥98% trace metals basis
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.999% trace metals basis