  • Search Results
  • Delayed-onset complications of facial soft tissue augmentation with permanent fillers in 85 patients.
Delayed-onset complications of facial soft tissue augmentation with permanent fillers in 85 patients.
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.] (2013-10-05)
Jonathan A Kadouch, Daniel J Kadouch, Shai Fortuin, Leo van Rozelaar, Refaat B Karim, Rick Hoekzema
ABSTRACT
To evaluate factors influencing the onset and type of adverse events in patients injected with permanent fillers in the face and to propose a therapeutic strategy for these complications. A prospectively attained series of 85 patients with delayed-onset complications after facial injection with permanent fillers underwent clinical follow-up and treatment of the complications. Lag times until onset and type of delayed-onset complication varied according to filler material. In 28% (n = 24) of the cases, patients reported the onset of complications after dental procedures, additional injections with fillers, or other invasive treatments in the facial area. Forty-eight (57%) patients required invasive treatment. Abscess formation was significantly more frequent in patients with human immunodeficiency virus infection and facial lipoatrophy (p = .001). The intrinsic characteristics of the injected filler and the immune status of the patient play important roles in the diversity of time of onset and type of delayed-onset adverse events observed. It seems that invasive facial or oral procedures in the vicinity of filler depots can provoke such complications. We propose a strategy for treating these complications and advise great caution when using permanent filling agents.
Sigma-Aldrich
Hyaluronic acid sodium salt from rooster comb, avian glycosaminoglycan polysaccharide
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus zooepidemicus, bacterial glycosaminoglycan polysaccharide
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, bacterial glycosaminoglycan polysaccharide
Sigma-Aldrich
Silicone oil, for oil baths (from −50°C to +200°C)
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 70,000-120,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 15,000-30,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 10,000-30,000
Sigma-Aldrich
Silicone oil, viscosity 5 cSt (25 °C)
Silicone oil, for melting point and boiling point apparatuses
Sigma-Aldrich
Silicone oil, viscosity 1,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 20 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 500 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 100 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 50 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 350 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 10 cSt (25 °C)
Sigma-Aldrich
Silicone oil, Dow Corning 200® fluid, viscosity 60,000 cSt (25 °C)
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,000,000-1,250,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,500,000-1,750,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 8,000-15,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 2,000,000-2,200,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,200
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 50,000-70,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 30,000-50,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 300,000-500,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 2,000,000-2,400,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 120,000-350,000
Sigma-Aldrich
Silicone oil, viscosity 30,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 100,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 10,000 cSt (25 °C)