  • Search Results
  • Fully-automated radiosynthesis and in vitro uptake investigation of [N-methyl-¹¹C]methylene blue.
Fully-automated radiosynthesis and in vitro uptake investigation of [N-methyl-¹¹C]methylene blue.
Anticancer research (2013-10-15)
Lutz F Schweiger, Tim A D Smith
ABSTRACT
Malignant melanoma is a type of skin cancer which can spread rapidly if not detected early and left untreated. Positron Emission Tomography (PET) is a powerful imaging technique for detecting cancer but with only a limited number of radiotracers available the development of novel PET probes for detection and prevention of cancer is imperative. In the present study we present the fully-automated radiosynthesis of [N-methyl-(11)C]methylene blue and an in vitro uptake study in metastasic melanoma cell lines. Using the GE TRACERlab FXc Pro module [N-methyl-(11)C]methylene blue was isolated via solid-phase extraction in an average time of 36 min after end of bombardment and formulated with a radiochemical purity greater than 95%. The in vitro uptake study of [N-methyl-(11)C]methylene blue in SK-MEL28 melanin-expressing melanoma cell line demonstrated in site-specific binding of 51% promoting it as a promising melanoma PET imaging agent.
Sigma-Aldrich
Methylene Blue solution, for microscopy, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Methylene Blue solution, suitable for microbiology
Sigma-Aldrich
Methylene Blue solution, for microscopy