Investigation on the antibacterial micro-porous titanium with silver nano-particles.
Journal of nanoscience and nanotechnology (2013-11-20)
Wei Dong, Yingchun Zhu, Jingxian Zhang, Liqiang Lu, Chengjian Zhao, Lifeng Qin, Yingbin Li
RESUMO
Micro-porous titanium is coated with silver nanoparticles by using a simple chemical reduction method that exhibits excellent antibacterial ability. Scanning electron microscopy (SEM) shows that the silver nanoparticles with average sizes of about 100 nm are formed homogeneously on the micro-porous titanium surface. After the micro-porous Ti coated with silver nano particles is treated by heating, the average size of the silver nano particles is slightly increased, but the nano particles are more uniformly dispersed on the surface of the micro-porous titanium. X-ray diffraction (XRD) indicates that those nanoparticles are metallic silver produced on the micro-porous titanium surface. The samples of micro-porous titanium coated with silver nanoparticles inhibit the growth of Escherichia coli. Our results show that the electrical double layer of the samples play an important role in the antibiosis and this study opens a new window for antibacterial mechanism which may be suitable for the other antibacterial metallic materials.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Silver, conductive paste
Sigma-Aldrich
Silver, nanopowder, <100 nm particle size, contains PVP as dispersant, 99.5% trace metals basis
Sigma-Aldrich
Silver, powder, 2-3.5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Silver nanowires, diam. × L 60 nm × 10 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Silver, flakes, 10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Silver, nanopowder, <150 nm particle size, 99% trace metals basis
Sigma-Aldrich
Titanium, powder, <45 μm avg. part. size, 99.98% trace metals basis
Sigma-Aldrich
Silver, powder, 5-8 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Titanium, powder, −100 mesh, 99.7% trace metals basis
Sigma-Aldrich
Silver nanowires, diam. × L 115 nm × 20-50 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Silver, colloidal, 65-75% Ag basis
Sigma-Aldrich
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Silver, shot, 1-3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, sponge, 3-19 mm, 99.5% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Titanium, nanoparticles, dispersion, <100 nm particle size, in mineral oil, 98.5% trace metals basis
Sigma-Aldrich
Titanium, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.995% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Sigma-Aldrich
Silver, powder, <45 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.81 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Silver, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, crystalline, 5-10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Sigma-Aldrich
Titanium, evaporation slug, diam. × L 6.3 mm × 6.3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, ≥99.99% trace metals basis