Genotoxicity of silver and titanium dioxide nanoparticles in bone marrow cells of rats in vivo.
Toxicology (2013-12-11)
Małgorzata M Dobrzyńska, Aneta Gajowik, Joanna Radzikowska, Anna Lankoff, Maria Dušinská, Marcin Kruszewski
RESUMO
Although nanomaterials have the potential to improve human life, their sideline effects on human health seem to be inevitable and still remain unknown. This study aimed to investigate the cytotoxicity and genotoxicity of titanium dioxide (TiO2) and silver (Ag) nanoparticles (NPs) at different doses and particle sizes to bone marrow cells. Both types of nanoparticles were chosen due to their wide applications of them in consumer products. Rats were injected intravenously with a single dose of 5 or 10 mg/kg bw of 20 nm AgNPs or with 5 mg/kg bw 200 nm AgNPs or with 5 mg/kg bw 21 nm TiO2NPs. The samples were taken at 24 h, 1 week and 4 weeks following the exposure. Micronucleus test and the Comet assay were used to detect DNA damage. Neither AgNPs nor TiO2NPs caused cytotoxicity to bone marrow red and white cells. The polychromatic erythrocytes are the main target of both nanoparticles. A single exposure to AgNPs induced significantly enhanced frequency of micronuclei not only at 24 h after exposure, but also 1 and 4 weeks later, whereas single exposure to TiO2NPs showed positive effect at 24 h only. Negative responses were shown in reticulocytes (micronuclei) and in leukocytes (Comet assay) of bone marrow. Results indicated that different bone marrow cells display different susceptibility toward genotoxicity mediated by both investigated nanoparticles. The use of materials containing nanoparticles and the potential health implication of them should be monitored.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Titanium(IV) oxide, nanopowder, 21 nm primary particle size (TEM), ≥99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, nanopowder, <25 nm particle size, 99.7% trace metals basis
Sigma-Aldrich
Silver, conductive paste
Sigma-Aldrich
Silver, nanopowder, <100 nm particle size, contains PVP as dispersant, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, 99.8% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, nanopowder, <100 nm particle size, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Silver, powder, 2-3.5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5%
Sigma-Aldrich
Silver nanowires, diam. × L 60 nm × 10 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Silver, flakes, 10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Silver, nanopowder, <150 nm particle size, 99% trace metals basis
Sigma-Aldrich
Titanium, powder, <45 μm avg. part. size, 99.98% trace metals basis
Sigma-Aldrich
Silver, powder, 5-8 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Titanium, powder, −100 mesh, 99.7% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Sigma-Aldrich
Silver nanowires, diam. × L 115 nm × 20-50 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Titanium(IV) oxide, rutile, 99.995% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.25 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
Sigma-Aldrich
Silver, colloidal, 65-75% Ag basis
Sigma-Aldrich
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis