Lead exposure among five distinct occupational groups: a comparative study.
Pakistan journal of pharmaceutical sciences (2014-01-01)
Mohammad Younis Gharaibeh, Karem Hasan Alzoubi, Omar Falah Khabour, Yousef Saleh Khader, Mamoun Abdallah Gharaibeh, Sulaiman Khalid Matarneh
RESUMO
This study was conducted to evaluate blood lead concentration among five selected occupational groups. The five groups were: hospital health workers, shop workers, taxi drivers, automobiles mechanics, and wood workers. The groups did not significantly differ among each other in the average of age and work years. ANOVA test revealed significantly higher mean lead blood concentration in taxi drivers, automechanics, and wood workers compared to other groups. Additionally, workers with lead concentration >0.483 umol/L (10μg/dL) were more likely to have frequent muscle pain compared to those with lower concentrations. No association between other symptoms of lead exposure/toxicity and blood lead concentration was detected. In conclusion, special attention must be directed toward lead blood levels and lead poisoning symptoms when examining patients from certain occupational groups such as taxi drivers, automechanics, and wood workers. Special safety precautions and educational programs are also needed to limit the lead exposure in these occupational groups.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Lead, powder, −100 mesh, 99.95% trace metals basis
Sigma-Aldrich
Lead, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Lead, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Lead, shot, <2 mm, 99.9% trace metals basis
Sigma-Aldrich
Lead, shot, 1-3 mm, 99.995% trace metals basis
Supelco
Lead (impurities), manufactured by MBH Analytical Ltd (83X PR8)