• Início
  • Resultados da busca
  • Role of insulin-like growth factor II receptor in transdifferentiation of free silica-induced primary rat lung fibroblasts.
Role of insulin-like growth factor II receptor in transdifferentiation of free silica-induced primary rat lung fibroblasts.
Biomedical and environmental sciences : BES (2014-01-08)
Chang Fu Hao, Xiao Fang Li, Wu Yao
RESUMO
To study the role of insulin-like growth factor II receptor in free silica-induced transdifferentiation of primary rat lung fibroblasts. Rat lung fibroblasts and rat alveolar macrophages were cultured. A transdifferentiation model of primary rat lung fibroblasts was induced by free silica. Levels of α-SMA protein, IGF-IIR protein and mRNA were measured by immunocytochemistry, Western blot and RT-PCR, respectively. Lung fibroblasts were treated with Wortmannin. The expression levels of α-SMA and IGF-IIR increased with the increasing free silica concentration and decreased after Wortmannin was used. The IGF-IIR plays an important role in free silica-induced transdifferentiation of primary rat lung fibroblasts.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
Sigma-Aldrich
LUDOX® TM-50 colloidal silica, 50 wt. % suspension in H2O
Sigma-Aldrich
Silicon dioxide, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
Sigma-Aldrich
Silica, mesostructured, MCM-41 type (hexagonal)
Sigma-Aldrich
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silica, nanopowder, spec. surface area 175-225 m2/g (BET), 99.8% trace metals basis
Sigma-Aldrich
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TMA colloidal silica, 34 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silica
Sigma-Aldrich
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silica, mesostructured, MSU-F (cellular foam)
Sigma-Aldrich
LUDOX® CL-X colloidal silica, 45 wt. % suspension in H2O
Sigma-Aldrich
Silicon dioxide, alumina doped, nanoparticles, dispersion, <50 nm particle size, 20 wt. % in H2O, 99.99% trace metals basis